Ultra Low Temperature Circulating Baths
Thermo Scientific™ ARCTIC A10 Refrigerated Circulators Available on GSA/VA Contract for Federal Government customers only.
A10 refrigerated bath circulators provide efficient cooling for open and closed applications, with temperatures ranging from −10° to +100°C and 6L maximum bath volume.
Thermo Scientific™ ARCTIC A25B Refrigerated Circulators Available on GSA/VA Contract for Federal Government customers only.
A25B refrigerated bath features a large work area, enabling high throughput and workflow efficiency, with temperatures ranging from −25° to +200°C and 21L maximum bath volume.
Thermo Scientific™ ARCTIC A40 Refrigerated Circulator Available on GSA/VA Contract for Federal Government customers only.
A40 refrigerated bath circulator provides efficient cooling for open and closed applications, with temperatures ranging from −40° to +200°C and 12L maximum bath volume.
Thermo Scientific™ ARCTIC A25 Refrigerated Circulators Available on GSA/VA Contract for Federal Government customers only.
A25 refrigerated bath circulator provides efficient cooling for open and closed applications, with temperatures ranging from −25° to +200°C and 12L maximum bath volume.
Thermo Scientific™ ARCTIC A10B Refrigerated Circulators Available on GSA/VA Contract for Federal Government customers only.
A10B refrigerated bath features a large work area, enabling high throughput and workflow efficiency, with temperatures ranging from −10° to +100°C and 30L maximum bath volume.
Thermo Scientific™ GLACIER Series G50 Ultra-Low Refrigerated Circulators Available on GSA/VA Contract for Federal Government customers only.
These Ultra-Low Refrigerated Circulators are cost effective with extreme temperature performance, offering high heating and cooling capacities for rapid heat-up and cool-down.
Thermo Scientific VersaCool™ Refrigerated Circulating Bath, -20° to +150°C Available on GSA/VA Contract for Federal Government customers only.
Advancements in remote monitoring capabilities, energy efficiency, global voltage input, sample capacity, and other tools and features give you control.
Thermo Scientific™ Ultra-Low Temperature Circulators Available on GSA/VA Contract for Federal Government customers only.
These Ultra-Low Temperature Circulators meet a variety of application needs. The ULT baths are designed primarily to circulate to external closed loop applications.
Heidolph™ Hei-Therm™ Refrigerated Circulator Bath Available on GSA/VA Contract for Federal Government customers only.
Intuitive large digital display with programmable time versus temperature ramps on AP models
Thermo Scientific™ ARCTIC A28 Refrigerated Circulator
A28 refrigerated bath circulator provides efficient cooling for open and closed applications, with temperatures ranging from −28° to +200°C and 10L maximum bath volume.
Thermo Scientific™ ARCTIC A5B Refrigerated Circulators
A5B refrigerated baths feature a large work area, enabling high throughput and workflow efficiency, with temperatures ranging from −5° to +100°C and 20L maximum bath volume.
Thermo Scientific™ ARCTIC A28F Refrigerated Circulator
A28F refrigerated bath circulator provides efficient cooling for open and closed applications, with temperatures ranging from −28° to +200°C and 10L maximum bath volume.
Thermo Scientific VersaCool™ Refrigerated Circulating Bath, -20° to +150°C: PROMO
Advancements in remote monitoring capabilities, energy efficiency, global voltage input, sample capacity, and other tools and features give you control.
Thermo Scientific™ Lindberg/Blue M™ Refrigerated Circulating Baths
These baths combine contemporary cabinet design, advanced microprocessor control and a range of accessories to suit a variety of laboratory or process applications.
Thermo Scientific™ ARCTIC A24B Refrigerated Circulators
A24B refrigerated bath features a large work area, enabling high throughput and workflow efficiency, with temperatures ranging from −24° to +200°C and 27L maximum bath volume.
Fisher Scientific™ Isotemp™ Refrigerating Circulators (220V)
Excellent temperature control for applications requiring refrigerated fluid work areas or pumping to an external application
  spinner