Centrifuge Rotors
Thermo Scientific™ 2000mL BIOS Swinging Bucket Rotors
The unique 8 x 2000mL (16L) capacity of the Thermo Scientific™ 2000mL BIOS swinging bucket rotors enhance the productivity of your bioprocessing methods.
Thermo Scientific™ S150-AT Fixed Angle Rotor Available on GSA/VA Contract for Federal Government customers only.
Perform high-speed applications with the Thermo Scientific™ S150-AT Fixed Angle Rotor, which operates at 150,000rpm with 8 x 2.0mL capacity.
Thermo Scientific™ HBB-6 Swinging Bucket Rotor Available on GSA/VA Contract for Federal Government customers only.
Swinging bucket rotor provides flexibility for blood banking
Thermo Scientific™ AH-629 (36mL) Swinging Bucket Rotor Available on GSA/VA Contract for Federal Government customers only.
Facilitate large volume particle isolations using Thermo Scientific™ AH-629 6 x 36mL Swinging-Bucket Rotor.
Thermo Scientific™ A22-24 x 16 Fixed Angle Rotor Available on GSA/VA Contract for Federal Government customers only.
Shorten run set-up time and gain peace-of-mind that the rotor is secure using the Thermo Scientific™ A22-24 x 16mL Fixed-Angle Rotor.
Thermo Scientific™ 1000mL BIOS Swinging Bucket Rotors
Efficiently and reliably perform your bioprocessing methods with the Thermo Scientific™ 6 x 1000mL or 8 x 1000mL swinging bucket rotors.
Thermo Scientific™ S120-AT2 Fixed Angle Rotor Available on GSA/VA Contract for Federal Government customers only.
Process large capacities (10 x 2mL) with this small, fixed angle rotor.
Thermo Scientific™ TX-200 Swinging Bucket Rotor Available on GSA/VA Contract for Federal Government customers only.
Accommodate a wide range of processing volumes with the TX-200 4 x 180mL Swinging Bucket Rotor.
Thermo Scientific™ AH-629 (17mL) Swinging Bucket Rotor Available on GSA/VA Contract for Federal Government customers only.
Process samples at 29,000rpm with the Thermo Scientific™ AH-629 6 x 17mL Swinging Bucket Rotor.
Thermo Scientific™ A23-6 x 100 Fixed Angle Rotor Available on GSA/VA Contract for Federal Government customers only.
Perform a variety of applications due to multiple adapters for smaller tubes with the Thermo Scientific™ A23-6x100 Fixed Angle Rotor.
Thermo Scientific™ StepSaver™ 70V6 Vertical Rotor Available on GSA/VA Contract for Federal Government customers only.
Minimize sample loss with the quick load and unload feature of the Thermo Scientific™ StepSaver™ 70V6 Vertical Rotor.
Thermo Scientific™ T-1270 Fixed Angle Titanium Rotor Available on GSA/VA Contract for Federal Government customers only.
Effectively perform a wide range of applications with the T-1270 Rotor, which offers 12-place capacity.
Thermo Scientific™ 8 x 50mL Individually Sealed Fixed Angle Rotor Available on GSA/VA Contract for Federal Government customers only.
Centrifuge eight conical tubes in individually sealed vessels.
Thermo Scientific™ HIGHConic™ Fixed-Angle Rotor Available on GSA/VA Contract for Federal Government customers only.
Process samples at a constant 45-degree angle with the Thermo Scientific™ HIGHConic™ Fixed-Angle Rotor.
Thermo Scientific™ TX-400 4 x 400mL Swinging Bucket Rotor Available on GSA/VA Contract for Federal Government customers only.
Support a wide range of sample sizes and applications with the Thermo Scientific™ TX-400 4 x 400mL Swinging Bucket Rotor.
Thermo Scientific™ Fiberlite™ F65L-6 x 13.5 Fixed-Angle Rotor Available on GSA/VA Contract for Federal Government customers only.
Process 6 x 13.5mL at speeds of 65,000rpm and maximum RCF of 324,000 x g with the corrosion-free carbon fiber Thermo Scientific™ Fiberlite™ F65L-6 x 13.5 Fixed-Angle Rotor.
Thermo Scientific™ SH-3000 Swinging Bucket Rotor Available on GSA/VA Contract for Federal Government customers only.
Process microplates, PCR plates/tube strips and many commonly used centrifugal filtration and purification devices with the Thermo Scientific™ SH-3000 Swinging Bucket Rotor.
Thermo Scientific™ Fiberlite™ F23-48 x 1.5 Fixed-Angle Rotor Available on GSA/VA Contract for Federal Government customers only.
Combine high g-force and high capacity for 1.5mL microtubes with the Thermo Scientific™ Fiberlite™ F23-48 x 1.5mL Fixed-Angle Rotor.
Thermo Scientific™ S120-AT3 Fixed Angle Rotor Available on GSA/VA Contract for Federal Government customers only.
Process large capacities (14 x 0.5mL) with this small, fixed angle rotor.
Thermo Scientific™ BIOLiner™ Swinging-Bucket Rotor Available on GSA/VA Contract for Federal Government customers only.
Spin samples at 3,500rpm with the Thermo Scientific™ BIOLiner™ Swinging-Bucket Rotor.
Thermo Scientific™ A27-6 x 50 Fixed Angle Rotor Available on GSA/VA Contract for Federal Government customers only.
Maintain low sample temperatures at all speeds while experiencing high RCF performance with the Thermo Scientific™ A27-6 × 50mL Fixed-Angle Rotor.
Thermo Scientific™ StepSaver™ 50V39 Vertical Rotor Available on GSA/VA Contract for Federal Government customers only.
Save time and improve accuracy of results with the titanium Thermo Scientific™ StepSaver™ 50V39 Vertical Rotor.
Thermo Scientific™ TX-100S Swinging Bucket Clinical Rotor Available on GSA/VA Contract for Federal Government customers only.
Provide efficient clinical processing of up to 8 x 5/7mL blood collection tubes with certified biocontainment using these rotors.
Thermo Scientific MicroClick 18 x 5 Fixed Angle Microtube Rotor Available on GSA/VA Contract for Federal Government customers only.
Reduce run times with high throughput 18-place capacity for 5.0mL microtubes and over 22,000 x g.
  spinner