Compact Rotors for Benchtop Centrifuges
Thermo Scientific MicroClick 18 x 5 Fixed Angle Microtube Rotor Available on GSA/VA Contract for Federal Government customers only.
Reduce run times with high throughput 18-place capacity for 5.0mL microtubes and over 22,000 x g.
Thermo Scientific™ 8 x 15mL Swinging Bucket Rotor Available on GSA/VA Contract for Federal Government customers only.
Ideal for centrifugation of blood collection tubes and medical laboratories or practices with low sample throughput
Thermo Scientific™ TX-100S Swinging Bucket Clinical Rotor Available on GSA/VA Contract for Federal Government customers only.
Provide efficient clinical processing of up to 8 x 5/7mL blood collection tubes with certified biocontainment using these rotors.
Thermo Scientific™ mySPIN™ Mini Centrifuge Series Rotors
Spin 0.2mL and 1.5/2.0mL tubes with Thermo Scientific™ mySPIN™ Mini Centrifuge Series Rotors.
Thermo Scientific™ MicroClick 30 x 2 Fixed Angle Microtube Rotor Available on GSA/VA Contract for Federal Government customers only.
Perform micro-volume applications such as nucleic acid preparation at up to 14,000rpm/21,693 x g with the Thermo Scientific™ MicroClick 30 x 2 Fixed Angle Microtube Rotor.
Thermo Scientific™ HIGHConic™ III Fixed Angle Rotor Available on GSA/VA Contract for Federal Government customers only.
View samples through a transparent lid for added safety and convenience in case of a spill.
Thermo Scientific™ CL10 4-Place Rotor Adapters Available on GSA/VA Contract for Federal Government customers only.
For use in Swinging Bucket Rotor 05-376-200
Thermo Scientific™ 6 x 50mL Fixed Angle HIGHConic™ Rotor with Cover Available on GSA/VA Contract for Federal Government customers only.
Enhance pelletization and reduce processing time in conical cell culture tubes with a unique 45° angle design by using the Thermo Scientific™ 6 × 50mL Fixed Angle Highconic™ Rotor.
Thermo Scientific™ TX-100 Swinging Bucket Clinical Rotor Available on GSA/VA Contract for Federal Government customers only.
Efficiently process up to 16 x 5/7mL blood collection tubes per run.
Thermo Scientific™ 8 x 8 PCR Strip Fixed Angle Rotor Available on GSA/VA Contract for Federal Government customers only.
Support micro-volume protocols, such as PCR reaction set-up with this 8 x 8 PCR centrifuge rotor, capable of spinning up to 14,800rpm/ 16,800 x g.
Thermo Scientific™ TX-150 Swinging Bucket Rotor Available on GSA/VA Contract for Federal Government customers only.
Perform a a wide range of clinical and research applications with the Thermo Scientific™ TX-150 Swinging Bucket Rotor, which is highly configurable and convenient.
Thermo Scientific™ MicroClick 24 x 2 Fixed Angle Microtube Rotor
Perform micro-volume applications such as nucleic acid preparation at up to 16,000rpm/ 24,372 x g with the Thermo Scientific™ MicroClick 24 x 2 microtube rotor.
Thermo Scientific™ TX-150 Swinging Bucket Rotor Available on GSA/VA Contract for Federal Government customers only.
Accomodates up to 24 x 5/7mL blood collection tubes or 8 x 50mL conical tubes at 4,500rpm/ 3,260 x g
Thermo Scientific™ 4 x 100mL Swinging Bucket Rotor Available on GSA/VA Contract for Federal Government customers only.
Combine Thermo Scientific™ 4 x 100mL Swinging Bucket Rotor with an optional aerosol-tight cap for added sample protection.
Thermo Scientific™ 4 x 180mL Swinging Bucket Rotor Available on GSA/VA Contract for Federal Government customers only.
Incorporate the Thermo Scientific™ 4 × 180mL Swinging Bucket Rotor with Sorvall™ Stratos™ rotors and adapters.
Thermo Scientific™ DualSpin™ Rotor
Perform both routine clinical and life science separations with this unique rotor featuring a 2-in-1 hybrid design.
Thermo Scientific™ MT-12 Microtube Swinging Bucket Rotor Available on GSA/VA Contract for Federal Government customers only.
Spin up to 12 microtubes and filtration columns up to 13,000rpm / 16,438 x g for small-volume research applications.
Thermo Scientific™ M10 Microplate Swinging Bucket Rotor Available on GSA/VA Contract for Federal Government customers only.
Spin up to 4 microplates or 2 deepwell plates with the convenience of transportable microplate carriers.
Thermo Scientific™ 24 x 1.5/2.0mL Drum Rotor Available on GSA/VA Contract for Federal Government customers only.
For Biofuge Stratos centrifuges
Thermo Scientific™ 236 Aerocarrier Rotor Available on GSA/VA Contract for Federal Government customers only.
Combine Thermo Scientific™ 236 Aerocarrier Rotor with CL2 rotors.
Thermo Scientific™ Hematocrit Fixed-Angle Rotor Available on GSA/VA Contract for Federal Government customers only.
Save time in clinical processing applications with a fixed-angle rotor that provides dedicated blood processing to achieve 24 capillary separations per run.
Thermo Scientific™ 16 x 16 Fixed Angle Rotor
For use in Heraeus Biofuge Stratos centrifuges
Thermo Scientific™ 8 x 50mL Fixed Angle Rotor Available on GSA/VA Contract for Federal Government customers only.
Carry out high-risk applications with the Thermo Scientific™ 8 x 50mL Fixed-Angle Rotor.
Thermo Scientific™ 6 x 94mL Fixed Angle Rotor Available on GSA/VA Contract for Federal Government customers only.
Process samples at 10,500/12,000rpm with the Thermo Scientific™ 6 x 94mL Fixed Angle Rotor.
Thermo Scientific™ 24 x 1.5/2.0mL Fixed Angle Rotor Available on GSA/VA Contract for Federal Government customers only.
Biocontainment-approved, autoclavable aluminum rotor with cover
Corning™ LSE™ Fixed-Angle Rotors Available on GSA/VA Contract for Federal Government customers only.
Rotors with different tube size and capacity configurations
  spinner