Psychrometers
EISCO Economy Sling Psychrometer Kit Products for Science Education
Easy to assemble kit to determine relative humidity.
H-B Instrument™ Durac™ Compact Sling Psychrometer Products for Science Education
Measures relative humidity with weather bureau-type wet and dry bulb.
H-B Instrument™ Durac™ Compact Sling Psychrometers
Designed with a built in slide rule to convert wet and dry bulb readings to relative humidity
Sling Psychrometer Kit Products for Science Education
Determine dew point and relative humidity.
Bacharach™ Replacement Parts for Sling Psychrometer
Replacement for wet- or dry-bulb thermometer in 11-664 Psychrometer
Bacharach™ Sling Psychrometer
No need to use tables or make calculations with this sling psychrometer
  spinner