Spectrometer Accessories
Alfa Aesar™ Disposable IR Cards, 5x10cm, KCl window, 9.5mm aperture
Provide a conveniently mounted, non-absorbing substrate for qualitative infrared analysis.
Thermo Scientific™ Crystal Plates and Spare Parts for the Thermo Scientific™ iD7 ATR Accessory
Choose the crystal plate to meet specific application and budget requirements
Alfa Aesar™ Disposable IR Cards, 5x10cm, NaCl window, 19mm aperture
Provide a conveniently mounted, non-absorbing substrate for qualitative infrared analysis.
Thermo Scientific™ Golden Gate ATR for Nicolet™ iS5 FT-IR Spectrometer
This versatile infrared sampling accessory is fast, sensitive, and robust
Alfa Aesar™ Disposable IR Cards, 5x10cm, NaCl window, 15mm aperture
Provide a conveniently mounted, non-absorbing substrate for qualitative infrared analysis.
Alfa Aesar™ Disposable IR Cards
Cardboard card that fits into the holder of a spectrophotometer
Alfa Aesar™ Disposable IR Cards, 5x10cm, NaCl window, 9.5mm aperture
Provide a conveniently mounted, non-absorbing substrate for qualitative infrared analysis.
Alfa Aesar™ Disposable IR Cards, 5x10cm, KBr window, 9.5mm aperture
Provide a conveniently mounted, non-absorbing substrate for qualitative infrared analysis.
Alfa Aesar™ Disposable IR Cards, 5x10cm, KCl window, 19mm aperture
Provide a conveniently mounted, non-absorbing substrate for qualitative infrared analysis.
Thermo Scientific™ iD5 ZnSe ATR for Nicolet™ iS5 FT-IR Spectrometer
High-performance singlebounce ZnSe ATR sample accessory for FTIR. Ideal for fast, reproducible sampling of solids, liquids, films, pastes, or gels.
Thermo Scientific™ Spectronic™ High-Resolution Spectral Libraries
Reference spectra to verfiy materials and identify unknown samples
Alfa Aesar™ Disposable IR Cards, 5x10cm, KBr window, 19mm aperture
Provide a conveniently mounted, non-absorbing substrate for qualitative infrared analysis.
Thermo Scientific™ Evolution™ Smart™ Sipper Accessories
Get the most out of your Thermo Scientific™ Evolution™ Spectrophotometer with a wide selection of Thermo Scientific™ Evolution™ Smart™ Sipper Accessories.
Bel-Art SP Scienceware™ Wilmad™ 4.0mm Solid State NMR Rotor Cap Remover
Eliminate possible damage of the cap and rotor with this zero turn cap remover. Bel-Art SP Scienceware™ Wilmad™ 4.0mm Solid State NMR Rotor Cap Remover simplifies removal of end caps and base plugs in solid state NMR.
Bel-Art SP Scienceware™ WIlmad™ OmniFit™ Valve NMR System Available on GSA/VA Contract for Federal Government customers only.
Designed for easy injection of chemicals through a gas-tight syringe for air sensitive samples without using a glove box. Bel-Art SP Scienceware™ WIlmad™ OmniFit™ Valve NMR System is comprised of a 5mm end cap, 2-way valve, septum with cap, barrel coupler and a vacuum connector.
Wilmad Luer Lock Syringe Needles Available on GSA/VA Contract for Federal Government customers only.
Designed for easy sample loading with Luer Lock syringes, these needles are available in stainless steel or PTFE. Bel-Art SP Scienceware™ Wilmad Luer Lock Syringe Needles are ideal for puncturing septum in air-sensitive samples.
Bel-Art SP Scienceware™ Wilmad Rotor and Caps for Bruker MAS Probe Available on GSA/VA Contract for Federal Government customers only.
Eliminate the need to solvate with MAS rotors manufactured from the highest purity Zirconia. Bel-Art SP Scienceware™ Wilmad MAS Rotors provide the ultimate solution for analysis of solid samples. Available for most present solid state NMR spectrometers.
Bel-Art SP Scienceware™ Wilmad™ Bottom Plug for 3.2mm MAS Rotor Available on GSA/VA Contract for Federal Government customers only.
Designed for use with MAS Rotors. Bel-Art SP Scienceware™ Wilmad™ Bottom Plug for 3.2mm MAS Rotor features a Vespal composition and works with applications ranging in temperature from -30° to 70°:C.
Bel-Art SP Scienceware™ Wilmad™ 150mL Suprasil Offset Nitrogen Dewar Flask for Bruker Available on GSA/VA Contract for Federal Government customers only.
Designed for cavities that have limited space between the reservoir and the drive shaft for iris. Bel-Art SP Scienceware™ Wilmad™ 150mL Suprasil Offset Nitrogen Dewar Flasks provide a convenient alternative to VT systems for studies at 77 K, and are silvered around the reservoir.
Bel-Art SP Scienceware™ Wilmad™ Pyrex™ MAS Rotor Inserts Available on GSA/VA Contract for Federal Government customers only.
Designed for air-sensitive samples and semi-solid samples such as gels or highly viscous liquids. Bel-Art SP Scienceware™ Wilmad™ Pyrex™ MAS Rotor Inserts allow samples to be sealed into an NMR rotor by heat-seal or epoxy.
Bel-Art SP Scienceware™ Wilmad™ NMR Tube pH Electrode Available on GSA/VA Contract for Federal Government customers only.
Measure pH range from 0 to 14 inside an NMR sample with this electrode. Bel-Art SP Scienceware™ Wilmad™ NMR Tube pH Electrodes are compatible with 5mm Thin Walled NMR tubes.
Thermo Scientific™ iD5 ATR Accessory for the Nicolet™ iS5
An ultra-high performance ATR accessory for FT-IR spectrometers
  spinner