Spectrometers Accessories
Thermo Scientific™ Evolution™ Smart™ Sipper Accessories
Get the most out of your Thermo Scientific™ Evolution™ Spectrophotometer with a wide selection of Thermo Scientific™ Evolution™ Smart™ Sipper Accessories.
Bel-Art SP Scienceware™ Wilmad Rotor and Caps for Bruker MAS Probe Available on GSA/VA Contract for Federal Government customers only.
Eliminate the need to solvate with MAS rotors manufactured from the highest purity Zirconia. Bel-Art SP Scienceware™ Wilmad MAS Rotors provide the ultimate solution for analysis of solid samples. Available for most present solid state NMR spectrometers.
Bel-Art SP Scienceware™ WIlmad™ OmniFit™ Valve NMR System Available on GSA/VA Contract for Federal Government customers only.
Designed for easy injection of chemicals through a gas-tight syringe for air sensitive samples without using a glove box. Bel-Art SP Scienceware™ WIlmad™ OmniFit™ Valve NMR System is comprised of a 5mm end cap, 2-way valve, septum with cap, barrel coupler and a vacuum connector.
Bel-Art SP Scienceware™ Wilmad™ Bottom Plug for 3.2mm MAS Rotor Available on GSA/VA Contract for Federal Government customers only.
Designed for use with MAS Rotors. Bel-Art SP Scienceware™ Wilmad™ Bottom Plug for 3.2mm MAS Rotor features a Vespal composition and works with applications ranging in temperature from -30° to 70°:C.
Bel-Art SP Scienceware™ Wilmad™ NMR Tube pH Electrode Available on GSA/VA Contract for Federal Government customers only.
Measure pH range from 0 to 14 inside an NMR sample with this electrode. Bel-Art SP Scienceware™ Wilmad™ NMR Tube pH Electrodes are compatible with 5mm Thin Walled NMR tubes.
Wilmad Luer Lock Syringe Needles Available on GSA/VA Contract for Federal Government customers only.
Designed for easy sample loading with Luer Lock syringes, these needles are available in stainless steel or PTFE. Bel-Art SP Scienceware™ Wilmad Luer Lock Syringe Needles are ideal for puncturing septum in air-sensitive samples.
Bel-Art SP Scienceware™ Wilmad™ 4.0mm Solid State NMR Rotor Cap Remover
Eliminate possible damage of the cap and rotor with this zero turn cap remover. Bel-Art SP Scienceware™ Wilmad™ 4.0mm Solid State NMR Rotor Cap Remover simplifies removal of end caps and base plugs in solid state NMR.
Bel-Art SP Scienceware™ Wilmad™ Pyrex™ MAS Rotor Inserts Available on GSA/VA Contract for Federal Government customers only.
Designed for air-sensitive samples and semi-solid samples such as gels or highly viscous liquids. Bel-Art SP Scienceware™ Wilmad™ Pyrex™ MAS Rotor Inserts allow samples to be sealed into an NMR rotor by heat-seal or epoxy.
Bel-Art SP Scienceware™ Wilmad™ 150mL Suprasil Offset Nitrogen Dewar Flask for Bruker Available on GSA/VA Contract for Federal Government customers only.
Designed for cavities that have limited space between the reservoir and the drive shaft for iris. Bel-Art SP Scienceware™ Wilmad™ 150mL Suprasil Offset Nitrogen Dewar Flasks provide a convenient alternative to VT systems for studies at 77 K, and are silvered around the reservoir.
SPEX™ SamplePrep Prep-Aid™ Grinding Aids Available on GSA/VA Contract for Federal Government customers only.
Ideal for samples which crumble or ablate after pressing.
Scientific Cell Glass Filter for Testing Wavelength and Photometric Accuracy
This glass filter is used for testing the wavelength accuracy and photometric accuracy (0.5 to 1 Abs).
SPEX CertiPrep™ Disposable XRF X-Cell™ Sample Cups Available on GSA/VA Contract for Federal Government customers only.
Permit fast XRF analysis of liquid samples.
SPEX™ SamplePrep Disposable XRF X-Cell™ Sample Cups Window Film Available on GSA/VA Contract for Federal Government customers only.
Strong and durable window film. SPEX SamplePrep Disposable XRF X-Cell Sample Cups Window Film covers the cell and serves as the sample window, allowing uniform transmission of X-rays.
SPEX™ SamplePrep Disposable XRF Spec-Cap™ Sample Cups Available on GSA/VA Contract for Federal Government customers only.
Aluminum cups aid in the production of pressed-powder sample discs for XRF.
Eppendorf™ Vis Cuvettes Available on GSA/VA Contract for Federal Government customers only.
Disposable cuvettes made of clear plastic with a light transmission of 300 nm to 900 nm. The Vis cuvettes are the perfect tool for applications outside of the UV range, for example, colorimetric protein assays (Bradford, Lowry, etc.), determining of the optical density of bacterial cultures (OD 600 methods), kinetic, and fluorescence measurements.
GE Healthcare Ultrospec 10 Disposable Cell
Disposable cell suited for use in OD600 measurements of bacterial cell cultures
  spinner