Spectrometers
3B Scientific™ Spectrometry Supplementary Set Products for Science Education
The 3B Scientific Spectrometry Supplementary Set is used for investigating spectrometry of transmitted signals and measurement of absorption losses.
EISCO Advanced Spectrometer Products for Science Education
This instrument whilst robust enough for individual student use and also intended for more advanced work.
Intermediate Spectrometer Products for Science Education
Robust enough for student use, yet capable of advanced work.
EISCO Basic Spectrometer Products for Science Education
An economical spectrometer for use in elementary schools to measure the angle of prisms using the law of reflection, demonstrate optics phenomenon with a diffraction grating or measure the wavelengths of monochromatic light sources.
Spectrometer Products for Science Education
Perform quantitative spectroscopy experiments using this good quality yet economical spectrometer.
Mott Manufacturing Mott Manufacturing Altus™ Table System Component, Female Ceiling Quick Connect Fitting
Mott quick connect fitting ensures internal piping and componens meet local codes and standards.
Otsuka America POCone™ Infrared Spectrophotometer
In vitro diagnostic device designed to measure changes in 13CO2 content in breath CO2 gas by infrared spectroscopic analysis
Public Lab Foldable Mini-Spectrometer Kit Products for Science Education
Visible/near-infrared spectrometer capable of connecting to smartphone.
Public Lab Desktop Spectrometry Kit Products for Science Education
Visible/near-infrared spectrometer capable of connecting to computer via USB cable.
American Educational Products Basic Spectrometer Products for Science Education
Rugged, all-metal spectrometer consists of an adjustable collimator tube, telescope and prism stand.
  spinner