Spectrophotometer Filters
Bel-Art™ SP Scienceware™ Klett™ Colorimeter Filters
Bel-Art™ SP Scienceware™ Klett™ Colorimeter Filters are glass filters for colorimetric and turbidimetric analyses to obtain readings on the Klett™ scale.
Hellma Analytics Potassium Chloride Filter
Liquid filter for testing stray light, according to Ph. Eur.
Hellma Analytics Sodium Iodide Filter
Liquid filter for testing stray light.
Hellma Analytics Ultrapure Water Filter
Reference filter for testing to stray light according to Ph. Eur.
Hellma Analytics Perchloric Acid
Reference filter for testing the photometric accuracy.
  spinner