Spectrophotometer Solutions and Reagents
Thermo Scientific™ NanoDrop™ One/OneC Productivity Kits
The Thermo Scientific™ NanoDrop™ One and NanoDrop™ OneC Productivity Kits allow you to improve the productivity of your NanoDrop One and NanoDrop OneC Microvolume UV-Vis Spectrophotometers.
Thermo Scientific™ NanoDrop™ CF-1 Calibration Fluid, 0.5mL ampule Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ NanoDrop CF-1 Calibration Fluid is an aqueous potassium dichromate (K2Cr207) solution for use in confirming spectrophotometer calibration.
Thermo Scientific™ Calibration Validation Carousels
Thermo Scientific™ Performance Verification Standards, Automated (CVC) will ensure the accuracy and reliability of your data while saving time and money.
Thermo Scientific™ NanoDrop™ PR-1 Reconditioning Kit Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ NanoDrop PR-1 Reconditioning Kit is used for reconditioning the sample measurement pedestals to a hydrophobic state if they become unconditioned.
Thermo Scientific™ NanoDrop™ CF-8 Calibration Check Kit Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ NanoDrop CF-8 Calibration Check Kit is an aqueous potassium dichromate (K2Cr2O7) solution for use in confirming spectrophotometer calibration.
Thermo Scientific™ PV-1 Performance Verification Solution Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ NanoDrop™ PV-1 Performance Verification Solution is used to help ensure the accuracy and reliability of your NanoDrop spectrophotometer data, helping you save time and reduce errors.
Thermo Scientific™ Calibration Validation Carousels Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Calibration Validation Carousels (CVC) will ensure the accuracy and reliability of your data while saving time and money.
Fisherbrand™ Secondary Spectrophotometric Calibration Standards
Calibration of the secondary standards is accomplished through the use of a high-performance UV/VIS/NIR spectrophotometer
LaMotte™ SMART™ Spectro Reagent Systems Products for Science Education Available on GSA/VA Contract for Federal Government customers only.
A wide range of reagent systems to meet your quality testing needs
EMD Millipore Total Dietary Fiber Reagant Kit Available on GSA/VA Contract for Federal Government customers only.
For determination of Total Dietary Fiber in foods.
EMD Millipore Spectroquant™ Surfactants (cationic) Cell Test Available on GSA/VA Contract for Federal Government customers only.
Combines higher sensitivity with easier handling, to ensure optimal results.
Lovibond™ Pocket Comparators DPD Tablets
For use with DPD Color Discs and Pocket Comparators
  spinner