Overhead Stirrers
Fisher Scientific™ CompactDigital™ Overhead Stirrer
Compact overhead stirrer, ideal for continuous, general purpose mixing. Easily fits into fume hoods or enclosed workstations.
Fisher Scientific™ Electric Analog Laboratory Overhead Stirrers: Model A15
Variable speed stirrers are electronically controlled
Fisher Scientific™ RealTorqueDigital™ Overhead Stirrers
High-torque, digital overhead stirrer offering speed and torque capabilities for a wide range of volumes and viscosities
Fisher Scientific™ Electric Analog Laboratory Overhead Stirrers: Model A40 Available on GSA/VA Contract for Federal Government customers only.
Variable speed stirrers are electronically controlled and built to handle viscosities ranging from light watery solutions to high viscosity fluids
Fisher Scientific™ Air-Powered Analog Overhead Stirrers
Variable speed and explosion-proof stirrers ideal for safely mixing all types of solvents, lacquers and volatile chemicals without the possiblity of spark ignition
Wheaton™ Overhead Stirrer Available on GSA/VA Contract for Federal Government customers only.
Ideal for most laboratory operations such as homogenizing or mixing
Fisher Scientific™ Electric Analog Laboratory Overhead Stirrers: Model A20 Available on GSA/VA Contract for Federal Government customers only.
Variable speed stirrers are electronically controlled and built to handle viscosities ranging from light watery solutions to high viscosity fluids
Fisher Scientific™ Electric Analog Laboratory Overhead Stirrers: Model A30
Variable speed stirrers are electronically controlled and built to handle viscosities ranging from light watery solutions to high viscosity fluids
Heidolph™ Radleys Tornado Overhead Stirring Station Available on GSA/VA Contract for Federal Government customers only.
Works exclusively with Carousel 6 Plus™ Reaction Stations for direct stirring from single overhead stirrer
Heidolph™ Mechanical Overhead Stirrer - RZR 1 Available on GSA/VA Contract for Federal Government customers only.
Unique overhead stirrer can be pivoted 90 degrees
Sper Scientific™ Minimixer Products for Science Education
Stir right in the test tube.
Caframo™ Real Torque Digital Stirrers
Three high-torque, digital overhead stirrers offering speed and torque capabilities for wide range of volumes and viscosities
Caframo™ CompactDigital™ Stirrer
Compact overhead stirrer, ideal for continuous, general purpose mixing. Easily fits into fume hoods or enclosed workstations.
Caframo™ Petite Digital Stirrer
Use this stirrer in formulation, R&D, clinical research and education labs, anywhere a magnetic stirrer is used. Caframo™ Petite Digital Stirrers are the ultimate in compact design measuring only two by four inches. Stirrer fits into small spaces and can be easily transported.
Caframo™ ReversingDigital™ Stirrer
With four working modes for complete mixing control
Caframo™ Petite Digital Stirrer
Complete, compact stirring package
Heidolph™ Mechanical Overhead Stirrers
Accomplish the most demanding tasks while providing the highest safety and increased performance life
Heidolph™ Electronic Overhead Stirrer - RZR 2051 Control
The powerful RZR stirrers accomplish the most demanding tasks while providing the highest safety and increased performance life
Heidolph™ Electronic Overhead Stirrer - RZR 2102 Control
2-gear stage stirrer which holds speed constant under significant load changes such as sticky media like a polymers
Heidolph™ Electronic Overhead Stirrer - RZR 2052 Control
Accepts torque of 180 Ncm in an overload situation and 90 Ncm for continuous operation at speeds from 30 to 1000rpm
  spinner