Stirrer Controllers
Thermo Scientific™ Cimarec™ 40M Controllers
Maintain constant speeds, even during viscosity changes with Thermo Scientific™ Cimarec™ 40M Controllers.
Thermo Scientific™ Cimarec™ i 20C Controller, 100-240V
Simplify operations with this user-friendly, menu-driven mircoprocessor controller included with the Micro, Mini, Compact, Maxi and Telesystems Stirrers.
Thermo Scientific™ Cimarec™ i Telemodul Controllers
Save space with Thermo Scientific™ Cimarec™ i Telemodul Controllers, including Micro, Mini, Compact and Maxi Stirrers.
Thermo Scientific™ Cimarec™ i 40C Controllers 100-240V
Receive higher speeds and more power settings than standard controllers by using Thermo Scientific™ Cimarec™ i 40C Controllers.
Thermo Scientific™ Cimarec™ 10M Controllers
Adjust speed from 100 to 1000rpm with this external control unit required for use with the Mobil 10 and Mobil 25 stirrers.
  spinner