Moveable Upright Cabinets
Labconco™ Protector™ Evidence Drying Cabinet: 5 ft.W
Provide a compact, space-saving method of securely drying evidence in your lab with the Labconco Protector Evidence Drying Cabinet. This durable, stainless steel-lined cabinet provides a secure ventilated area to dry evidence such as clothing and bedding and its HEPA and carbon filtered exhaust protects personnel from exposure to biohazards while eliminating nuisance odors.
Labconco™ Protector™ Benchtop Evidence Drying Cabinet
Provides a compact, space-saving method of securely drying evidence in a ventilated area
Mystaire™ Forensic Evidence Drying Cabinet
Safely dry wet evidence while protecting it from cross-contamination and workers from harmful fumes and odors
  spinner