Sound Apparatus
Guinea and Feather Apparatus Products for Science Education
Demonstrate the presence of a vacuum.
3B Scientific™ Resonance Ring Products for Science Education
Demonstrates vibration knots in determination of different frequencies.
3B Scientific™ Resonance Bowl Products for Science Education
Unique, historic way to demonstrate standing waves
3B Scientific™ Helmholtz Resonator Products for Science Education
Hollow glass bulb with a narrow tube leading out for demonstrating acoustic resonance.
3B Scientific™ Graduated Monochord Products for Science Education
Investigate the pitch of vibrating strings as a function of their tension, length, and thickness.
EISCO Tuning Forks, Set of Eight Products for Science Education
Demonstrate eight different frequencies with one set.
EISCO Melde's Apparatus Products for Science Education
Designed to show the effects of vibrations in a stretched cord, and investigate the relationship between frequency, tension and density.
3B Scientific™ Kundts Tube Products for Science Education
Demonstrate standing sound waves using this glass tube and calculate wavelengths of sound using a method devised by Kundt.
3B Scientific™ Electric Doorbell Products for Science Education
Demonstrates electro-magnetic operation and verifies that sound waves do not propagate in fine vacuum.
EISCO Resonant Tuning Fork Set Products for Science Education
Demonstrates acoustical resonance and sympathetic vibration.
3B Scientific™ Tuning Forks Set Products for Science Education
For experiments in acoustics
Eisco™ Tuning Fork Octave Set Products for Science Education
For teaching octaves
Tuning Forks Products for Science Education
Frequency and scale letter are stamped on each fork.
United Scientific Supplies Morse Code Experiment Kit Products for Science Education
Designed to show the principle of a telegraph.
Diatonic Scale Tuning Forks Products for Science Education
Harmonious chords familiar to piano players!
GSC Intenational Graduated Resonance Tube Products for Science Education
Choose GSC tubes for demonstrating resonance in the classroom.
American Educational Products Tune Tube Products for Science Education
Explore the science of sound.
Tuning Fork Activator Products for Science Education
No-hands activator for tuning forks.
Adjustable Tuning Fork Products for Science Education
Precision-grade aluminum alloy.
Tuning Fork Mallet Products for Science Education
Suitable for use on all tuning forks
GSC Resonance Apparatus
A simple apparatus that measures the velocity of sound in the air. The GSC Resonance Apparatus measures the velocity of sound in the air. Partially fill the resonance tube with water and then sound a tuning fork of known frequency at the tubes open upper end of the Resonance Tube.
Vibrating Tube Set Products for Science Education
Excellent apparatus for demonstrating vibration and beat phenomenon.
Doppler Effect Demo Kit Products for Science Education
Uses 256Hz, 512Hz and 1024Hz frequencies
GSC International Resonance Apparatus
Ideal for demonstration of standing waves. GSC International Resonace Apparatus can be used to observe resonance phenomena, speed of sound in air by exploiting standing waves, resonance effects in longitudinal waves, and resonance points for certain frequency and their relation with standing wavelength.
  spinner