Electrophoresis
Invitrogen™ iBlot™ 2 Transfer Stacks, nitrocellulose, regular size Greener Choice Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Used to transfer proteins with the iBlot 2 Gel Transfer Device
Thermo Scientific™ PageRuler™ Plus Prestained Protein Ladder, 10 to 250 kDa Available on GSA/VA Contract for Federal Government customers only.
Mixture of 9 blue-, orange- and green-stained proteins (10 to 250kDa) used as size standards in protein electrophoresis (SDS-PAGE) and Western blotting.
Thermo Scientific™ Owl™ EasyCast™ B2 Mini Gel Electrophoresis Systems Edge Available on GSA/VA Contract for Federal Government customers only.
Owl EasyCast B2 Mini Gel Electrophoresis Systems provide superior flexibility in a small footprint.
Thermo Scientific™ Owl™ EC300XL2 Compact Power Supply
Retain settings after shut-down with the power-off memory on the Thermo Scientific™ Owl™ EC300XL2 Compact Power Supply, ideal for DNA/RNA and Protein electrophoresis.
Invitrogen™ SimplyBlue™ SafeStain Greener Choice Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Less hazardous and does not require methanol or acetic acid fixatives or destains
Thermo Scientific™ Owl™ JGC Gel Casting Systems
Cast polyacrylamide gels faster with Thermo Scientific™ Owl™ JGC Gel Casting Systems, vertical protein gel casters that offer a unique casting method.
Thermo Scientific™ Pierce™ Silver Stain Kit
Stain proteins in 1D and 2D gels with this rapid, ultra-sensitive and versatile silver stain system that yields consistent and reliable results.
Invitrogen™ Bolt™ Empty Mini Gel Cassettes Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Allows users to pour custom gel formulations while still enjoying the advantages of the Mini Gel Tank
Invitrogen™ DryEase™ Mini-Gel Drying System Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
For use with DryEase™ Mini-Gel Drying System
Invitrogen™ XCell SureLock™ Mini-Cell Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Quickly and easily run up to two Novex 8 x 8cm minigels, leak-free without any clamps or grease
Invitrogen™ iBlot™ 2 Transfer Stacks, nitrocellulose, mini Greener Choice Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Used to transfer proteins with the iBlot 2 Gel Transfer Device
Thermo Scientific™ Owl™ EasyCast™ B1A Mini Gel Electrophoresis Systems Edge Available on GSA/VA Contract for Federal Government customers only.
Cast and run a gel in the buffer chamber and achieve flat, even banding patterns on a consistent basis with these B1A Mini Gel Electrophoresis systems.
Invitrogen™ Mini Gel Tank Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Compatible with a variety of Novex™ gels, NuPAGE™ gels, and Bolt™ Bis-Tris Plus gels
Invitrogen™ PowerEase™ 90W Power Supply (115 VAC) Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Designed specifically for mini-gel electrophoresis
Invitrogen™ SeeBlue™ Plus2 Pre-stained Protein Standard Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Allows you to easily visualize protein molecular weight ranges during electrophoresis and quickly evaluate Western transfer efficiency
Thermo Scientific™ Pierce™ Silver Stain for Mass Spectrometry
Reversibly stain proteins in polyacrylamide gels to identify bands to excise; then destain the gel pieces for analysis by mass spectrometry.
Invitrogen™ DryEase™ Mini Cellophane Non-distribution product as customer accommodation.
Component of DryEase Mini-Gel Drying System
Thermo Scientific™ Pierce™ Lane Marker Reducing Sample Buffer Available on GSA/VA Contract for Federal Government customers only.
Ready-to-use 5X SDS-PAGE sample loading buffers available in reducing and nonreducing formulations, both with a pink dye buffer-front marker.
Fisherbrand™ Electrophoresis Power Supplies Edge
Easy-to-use power supplies designed for safety
Invitrogen™ iBlot™ 2 Transfer Stacks, PVDF, regular size Greener Choice Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Consumable stacks with integrated PVDF transfer membranes for dry blotting of proteins
Cytiva (Formerly GE Healthcare Life Sciences) PlusOne™ Silver Staining Kit for Protein
Use for reproducible and sensitive staining of proteins in polyacrylamide gels. PlusOne™ Silver Staining Kit for Protein enables staining of proteins in less than two hours.
Thermo Scientific™ Owl™ EasyCast™ B1 Mini Gel Electrophoresis Systems Edge Available on GSA/VA Contract for Federal Government customers only.
Cast and run a gel in the same chamber with no tape or additional parts required with the Thermo Scientific™ Owl™ EasyCast™ B1 Mini Gel Electrophoresis Systems.
Invitrogen™ SilverQuest™ Silver Staining Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
30-fold more sensitive than staining with colloidal Coomassie™ G-250
Invitrogen™ Empty Gel Cassettes, mini, 1.5 mm Non-distribution product as customer accommodation.
Platic cassettes pre-sealed with slot pre-taped
Invitrogen™ 10X Bolt™ Sample Reducing Agent Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Optimized for use with Bolt™ Bis-Tris Plus gels, and are available in a variety of formats
Invitrogen™ StainEase™ Staining Trays Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Stains and solutions can be changed without having to handle the gel
Cytiva (Formerly GE Healthcare Life Sciences) miniVE Vertical Electrophoresis System Available on GSA/VA Contract for Federal Government customers only.
Performs 1-D and 2-D electrophoresis and electrotransfer in single compact unit
  spinner