Acrylamide Gels
Invitrogen™ Bolt™ 4 to 12%, Bis-Tris, 1.0 mm, Mini Protein Gel, 15-well Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Separation Range: 3.5 to 260 kd; Separation Type: Denaturing; For Use With: Mini Gel Tank, XCell SureLock Mini
Invitrogen™ Bolt™ 4 to 12%, Bis-Tris, 1.0 mm, Mini Protein Gel, 10-well Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Separation Range: 3.5 to 260 kd; Separation Type: Denaturing; For Use With: Mini Gel Tank, XCell SureLock Mini
Invitrogen™ NuPAGE™ 4 to 12%, Bis-Tris, 1.0 mm, Midi Protein Gel, 20-well Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Separation Range: 3.5 to 260 kd; Separation Type: Denaturing; For Use With: Midi Gel Tank, XCell4 SureLock Midi
Invitrogen™ NuPAGE™ 4 to 12%, Bis-Tris, 1.0 mm, Midi Protein Gel, 26-well Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Separation Range: 3.5 to 260 kd; Separation Type: Denaturing; For Use With: Midi Gel Tank, XCell4 SureLock Midi
Invitrogen™ Bolt™ 4 to 12%, Bis-Tris, 1.0 mm Mini Protein Gel, 12-well Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Separation Range: 3.5 to 260 kd; Separation Type: Denaturing; For Use With: Mini Gel Tank, XCell SureLock Mini
Invitrogen™ NuPAGE™ 4 to 12%, Bis-Tris, 1.0 mm, Mini Protein Gel, 2D-well Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Separation Range: 3.5 to 260 kd; Separation Type: Denaturing; For Use With: ZOOM IPGRunner Mini
Invitrogen™ NuPAGE™ 4 to 12%, Bis-Tris, 1.0 mm, Mini Protein Gel, 17-well Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Separation Range: 3.5 to 260 kd; Separation Type: Denaturing; For Use With: Mini Gel Tank, XCell SureLock Mini
Invitrogen™ Bolt™ 4 to 12%, Bis-Tris, 1.0 mm Mini Protein Gel, 17-well Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Separation Range: 3.5 to 260 kd; Separation Type: Denaturing; For Use With: Mini Gel Tank, XCell SureLock Mini
Invitrogen™ NuPAGE™ 10%, Bis-Tris, 1.5 mm, Mini Protein Gel, 10-well Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Separation Range: 3.5 to 260 kd; Separation Type: Denaturing; For Use With: Mini Gel Tank, XCell SureLock Mini
Invitrogen™ NuPAGE™ 4 to 12%, Bis-Tris, 1.0 mm, Midi Protein Gel, 12+2-well Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Separation Range: 3.5 to 260 kd; Separation Type: Denaturing; For Use With: Midi Gel Tank, XCell4 SureLock Midi
Invitrogen™ NuPAGE™ 10%, Bis-Tris, 1.0 mm, Mini Protein Gel, 15-well Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Separation Range: 3.5 to 260 kd; Separation Type: Denaturing; For Use With: Mini Gel Tank, XCell SureLock Mini
Invitrogen™ NuPAGE™ 3 to 8%, Tris-Acetate, 1.5 mm, Mini Protein Gel, 15-well Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Separation Range: 36 to 500 kd; Separation Type: Denaturing, Native; For Use With: Mini Gel Tank, XCell SureLock Mini
Invitrogen™ NativePAGE™ 4 to 16%, Bis-Tris, 1.0 mm, Mini Protein Gel, 10-well Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Separation Range: 30 to 10,000 kd; Separation Type: Native; For Use With: Mini Gel Tank, XCell SureLock Mini
Invitrogen™ NuPAGE™ 3 to 8%, Tris-Acetate, 1.5 mm, Mini Protein Gel, 10-well Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Separation Range: 36 to 500 kd; Separation Type: Denaturing, Native; For Use With: Mini Gel Tank, XCell SureLock Mini
Invitrogen™ NuPAGE™ 12%, Bis-Tris, 1.0 mm, Mini Protein Gel, 15-well Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Separation Range: 3.5 to 80 kd; Separation Type: Denaturing; For Use With: Mini Gel Tank, XCell SureLock Mini
Invitrogen™ Novex™ 10 to 20%, Tricine, 1.0 mm, Mini Protein Gel, 15-well Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Separation Range: 2.5 to 200 kd; Separation Type: Denaturing; For Use With: Mini Gel Tank, XCell SureLock Mini
Invitrogen™ NativePAGE™ 3 to 12%, Bis-Tris, 1.0 mm, Mini Protein Gel, 10-well Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Separation Range: 30 to 10,000 kd; Separation Type: Native; For Use With: Mini Gel Tank, XCell SureLock Mini
Invitrogen™ NuPAGE™ 10%, Bis-Tris, 1.5 mm, Mini Protein Gel, 15-well Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Separation Range: 3.5 to 260 kd; Separation Type: Denaturing; For Use With: Mini Gel Tank, XCell SureLock Mini
Invitrogen™ NuPAGE™ 10%, Bis-Tris, 1.0 mm, Midi Protein Gel, 26-well Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Separation Range: 3.5 to 260 kd; Separation Type: Denaturing; For Use With: Midi Gel Tank, XCell4 SureLock Midi
Invitrogen™ Novex™ 10 to 20%, Tricine, 1.0 mm, Mini Protein Gel, 10-well Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Separation Range: 2.5 to 200 kd; Separation Type: Denaturing; For Use With: Mini Gel Tank, XCell SureLock Mini
Invitrogen™ NuPAGE™ 3 to 8%, Tris-Acetate, 1.0 mm, Midi Protein Gel, 26-well Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Separation Range: 36 to 500 kd; Separation Type: Denaturing, Native; For Use With: Midi Gel Tank, XCell4 SureLock Midi
Invitrogen™ Novex™ 10 to 20%, Tricine, 1.0 mm, Mini Protein Gel, 12-well Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Separation Range: 2.5 to 200 kd; Separation Type: Denaturing; For Use With: Mini Gel Tank, XCell SureLock Mini
Invitrogen™ Bolt™ 10%, Bis-Tris, 1.0 mm, Mini Protein Gel, 12-well Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Separation Range: 3.5 to 260 kd; Separation Type: Denaturing; For Use With: Mini Gel Tank, XCell SureLock Mini
Invitrogen™ Novex™ 16%, Tricine, 1.0 mm, Mini Protein Gel, 10-well Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Separation Range: 2.5 to 200 kd; Separation Type: Denaturing; For Use With: Mini Gel Tank, XCell SureLock Mini
Invitrogen™ NativePAGE™ 4 to 16%, Bis-Tris, 1.0 mm, Mini Protein Gel, 15-well Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Separation Range: 30 to 10,000 kd; Separation Type: Native; For Use With: Mini Gel Tank, XCell SureLock Mini
Invitrogen™ NativePAGE™ 3 to 12%, Bis-Tris, 1.0 mm, Mini Protein Gel, 15-well Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Separation Range: 30 to 10,000 kd; Separation Type: Native; For Use With: Mini Gel Tank, XCell SureLock Mini
Invitrogen™ E-PAGE™ 8%, 3.7 mm, Midi Protein Gel, 48-well Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Separation Range: 20 to 100 kd; Separation Type: Denaturing; For Use With: E-BASE™
Invitrogen™ Bolt™ 8%, Bis-Tris, 1.0 mm, Mini Protein Gel, 15-well Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Separation Range: 3.5 to 260 kd; Separation Type: Denaturing; For Use With: Mini Gel Tank, XCell SureLock Mini
  spinner