Silver Stains for Protein Gels
Thermo Scientific™ Pierce™ Silver Stain Kit
Stain proteins in 1D and 2D gels with this rapid, ultra-sensitive and versatile silver stain system that yields consistent and reliable results.
Thermo Scientific™ Pierce™ Silver Stain for Mass Spectrometry
Reversibly stain proteins in polyacrylamide gels to identify bands to excise; then destain the gel pieces for analysis by mass spectrometry.
Cytiva (Formerly GE Healthcare Life Sciences) PlusOne™ Silver Staining Kit for Protein
Use for reproducible and sensitive staining of proteins in polyacrylamide gels. PlusOne™ Silver Staining Kit for Protein enables staining of proteins in less than two hours.
Thermo Scientific™ Pierce™ Color Silver Stain Kit
Detect proteins with this metallic silver staining technique that produces color variations to differentiate protein bands and spots in 1D and 2D gels.
Thermo Scientific™ Pierce™ Silver Stain Rescue Reagent
Remove undesirable background from unevenly and overly developed silver-stained polyacrylamide electrophoresis gels.
MP Biomedicals™ Silver Enhancing Kit, 2x250mL
Used for amplifying the signal from gold particles attached to proteins or antigens immobilized on membranes
  spinner