ELISA Reagents, Plates and Accessories
Alfa Aesar™ Mouse Osteocalcin Enzyme Immunoassay Kit
For measuring levels of osteocalcin in mouse serum and plasma samples
R&D Systems™ Human IL-5 Quantikine ELISA Kit, R&D Systems™
a 4.5 hour solid phase ELISA designed to measure IL-5 levels in cell culture supernates, serum, plasma, and urine.
R&D Systems™ Mouse IL-4 Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 4.5 hour solid phase ELISA designed to measure mouse IL-4 levels in cell culture supernates and serum.
R&D Systems™ Human CXCL8/IL-8 Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 4.5 hour solid-phase ELISA designed to measure human SDF-1 alpha in cell culture supernates and platelet-poor plasma.
Thermo Scientific™ Pierce™ High Binding Streptavidin Coated Plates
Bind biotinylated oligonucleotides and peptides with higher binding efficiency than other commercially available streptavidin-coated plates.
R&D Systems™ Human IL-6 DuoSet, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
contains the basic components required for the development of sandwich ELISAs to measure natural and recombinant Human IL-6
R&D Systems™ Human IL-1 alpha/IL-1F1 Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 3.5 or 4.5 hour solid phase ELISA designed to measure IL-1 alpha in cell culture supernates, serum, and plasma.
Invitrogen™ eBioscience™ Foxp3 / Transcription Factor Staining Buffer Set Non-distribution product as customer accommodation.
Formulated and optimized for staining with antibodies against transcription factors and nuclear proteins (e.g. Foxp3 and Ki-67), cytokines and chemokines
R&D Systems™ Human CD25/IL-2 R alpha Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
A 3.5 hour solid phase ELISA designed to measure IL-2 sR alpha in cell culture supernates, serum, and plasma. It contains NS0-expressed recombinant human IL-2 sR alpha and antibodies raised against the recombinant factor.
Invitrogen™ Novex™ Invitrogen™ IgG Subclass Human ELISA Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
For use with immunoglobulin and antibody isotyping
R&D Systems™ Human Lipocalin-2/NGAL Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 4.5 hour solid-phase ELISA designed to measure human Lipocalin-2 in cell culture supernates, serum, plasma, urine, and saliva.
R&D Systems™ Mouse IL-12 p70 Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 4.5 hour solid phase ELISA designed to measure mouse IL-12 p70 in cell culture supernates and mouse serum.
R&D Systems™ DuoSet Ancillary Reagent Kit 2 Available on GSA/VA Contract for Federal Government customers only.
Sufficient for at least five 96-well plates
R&D Systems™ Human GM-CSF Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
A 3.5 or 4.5 hour solid phase ELISA designed to measure GM-CSF levels in cell culture supernates, serum, and plasma. It contains E. coli-expressed recombinant human GM-CSF and antibodies raised against the recombinant factor.
R&D Systems™ Human VEGF Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
A 4.5 hour solid phase ELISA designed to measure VEGF165 levels in cell culture supernates, serum, and plasma. It contains Sf 21-expressed recombinant human VEGF165 and antibodies raised against the recombinant protein.
Thermo Scientific™ Pierce™ TMB Substrates Available on GSA/VA Contract for Federal Government customers only.
Horseradish peroxidase (HRP) substrates for ELISA that change from blue to yellow upon addition of a sulfuric or phosphoric acid stop solution.
R&D Systems™ Human CCL2/MCP-1 Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 3.5 or 4.5 hour solid phase ELISA designed to measure MCP-1 in cell culture supernates, serum, plasma, and urine.
R&D Systems™ Human IL-10 Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 3.5 or 4.5 hour solid phase ELISA designed to measure IL-10 in cell culture supernates, serum, and plasma.
Novus Biologicals™ Human HMGB1/HMG-1 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
R&D Systems™ Human sICAM-1/CD54 Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 2.0 hour solid phase ELISA designed to measure sICAM-1 in cell culture supernates, serum, and plasma.
R&D Systems™ Human IL-1 beta/IL-1F2 Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 3.5 or 4.5 hour solid phase ELISA designed to measure IL-1 beta in cell culture supernates, serum, and plasma.
R&D Systems™ Human CCL5/RANTES DuoSet ELISA Kit
Optimized capture and detection antibody pairings with recommended concentrations save lengthy development time
R&D Systems™ Mouse IL-13 Culture Media Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 4.5 hour solid phase ELISA designed to measure mouse IL-13 levels in cell culture supernates.
Invitrogen™ Mouse EGF ELISA Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Assay mouse EGF levels in cell culture supernatants, plasma, serum samples.
R&D Systems™ Human IL-16 Quantikine ELISA Kit, R&D Systems™
a 4.5 hour solid phase ELISA designed to measure human IL-16 in cell culture supernates, serum, and plasma.
R&D Systems™ Mouse CCL3/MIP-1 alpha Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 4.5 hour solid phase ELISA designed to measure mouse MIP-1 alpha levels in cell culture supernates and serum.
R&D Systems™ Human IL-2 Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
A 4.5 hour solid phase ELISA designed to measure IL-2 levels in cell culture supernates, serum, and plasma. It contains E. coli-expressed recombinant human IL-2 and antibodies raised against recombinant human IL-2.
R&D Systems™ Mouse IL-6 Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 4.5 hour solid-phase ELISA designed to measure mouse IL-6 in cell culture supernates, serum, and plasma.
  spinner