1-B ELISA Kits
Invitrogen™ Aldosterone Competitive ELISA Kit Non-distribution product as customer accommodation.
Assay Aldosterone levels in Extracted Serum, EDTA or Heparin Plasma, and Urine, Fecal Extracts, and Tissue Culture Media.
R&D Systems™ Total BDNF Quantikine ELISA Kit Available on GSA/VA Contract for Federal Government customers only.
For the quantitative determination of free- and Trk-bound Brain-Derived Neurotrophic Factor (Total BDNF). The Quantikine Total BDNF Immunoassay is a 3.5 hour solid phase ELISA designed to measure BDNF in cell culture supernates, tissue lysates, serum, plasma, urine, and human milk.
Invitrogen™ Mouse Axl ELISA Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Assay mouse Axl levels in cell culture supernatants, plasma, serum samples.
Invitrogen™ eBioscience™ Human s90K/Mac-2BP Platinum ELISA Kit Non-distribution product as customer accommodation.
For the quantitative detection of soluble human 90K/Mac-2 Binding Protein.
R&D Systems™ Human APRIL/TNFSF13 DuoSet, 15 Plate
Sufficient for at least 15 96-well plates
Novus Biologicals™ Human Activin B/Inhibin beta B ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
R&D Systems™ Mouse alpha-Fetoprotein/AFP Quantikine ELISA Kit
a 4.5 hour solid phase ELISA designed to measure mouse AFP levels in cell culture supernates, serum, and plasma.
Invitrogen™ Human Albumin (ALB) ELISA Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Assay human Albumin (ALB) levels in cell culture supernatants, plasma, serum samples.
Invitrogen™ Rat BDNF ELISA Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Assay rat BDNF levels in cell culture supernatants, plasma, serum samples.
Invitrogen™ Human Apolipoprotein C1 ELISA Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Assay human Apolipoprotein C1 levels in cell culture supernatants, plasma, serum samples.
Abnova™ NT5E (Human) ELISA Kit
Use to measure quantitatively human NT5E. Sandwich enzyme immunoassay.
R&D Systems™ Human Amyloid beta (aa1-42) Quantikine ELISA Kit
For the quantitative determination of human Amyloid beta (amino acids 1-40) concentrations in cell culture supernates, tissue lysates, and cerebrospinal fluid. Human Amyloid beta (aa1-42) Quantikine ELISA Kit employs the quantitative sandwich enzyme immunoassay technique.
R&D Systems™ Human Amyloid beta (aa1-40) Quantikine ELISA Kit
For the quantitative determination of human Amyloid beta (amino acids 1-40) concentrations in cell culture supernates, tissue lysates, and cerebrospinal fluid. Human Amyloid beta (aa1-40) Quantikine ELISA Kit employs the quantitative sandwich enzyme immunoassay technique.
Invitrogen™ eBioscience™ Human BAFF Instant ELISA Kit Non-distribution product as customer accommodation.
For the quantitative detection of human BAFF in cell culture supernatants, human serum, plasma, or other body fluids
Invitrogen™ eBioscience™ phospho-AKT 1/2/3 (Ser473) InstantOne™ ELISA Kit Non-distribution product as customer accommodation.
Specifically engineered for accurate measurement of phosphorylated human, mouse, and hamster AKT 1/2/3 in cell lysates. Detection of rat AKT 1/2/3 is expected. The InstantOne™ ELISA kit allows for analysis of samples in approximately one hour.
eBioscience™ Mouse Granzyme B ELISA Ready-SET-Go!™ Kit Non-distribution product as customer accommodation.
Specifically engineered for accurate and precise measurement of Mouse Granzyme B protein levels in serum, plasma, and tissue culture supernant samples.
Invitrogen™ Human Alpha-fetoprotein ELISA Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Assay human Alpha-fetoprotein (AFP) levels in cell culture supernatants, plasma, serum samples.
eBioscience™ Human Bcl-2 Platinum ELISA Kit Non-distribution product as customer accommodation.
For the quantitative detection of human Bcl-2.
Invitrogen™ Mouse BLC (CXCL13) ELISA Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Assay mouse BLC (CXCL13) levels in cell culture supernatants, plasma, serum samples.
R&D Systems™ Rat Adiponectin/Acrp30 DuoSet, 5 Plate
Sufficient for at least five 96-well plates
R&D Systems™ Human Arresten DuoSet ELISA
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Abnova™ ASCA IgG ELISA Kit
Enzyme-linked immunosorbent assay for the detection of IgG antibodies to Saccharomyces cerevisiae.
Novus Biologicals™ Human Adenylate Cyclase 3 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Novus Biologicals™ Non-species specific Apoptosis Detection ELISA ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
R&D Systems™ Human ADAMTS5 DuoSet ELISA Kit
This Human ADAMTS5 DuoSet ELISA Development kit contains the basic components required for the development of sandwich ELISAs to measure natural and recombinant human A Disintegrin and Metalloproteinase with Thrombospondin Motifs 5 (ADAMTS5).
Novus Biologicals™ Human Arg8-Vasopressin ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Novus Biologicals™ Human APP ELISA Kit (Chemiluminescence)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Novus Biologicals™ Bovine Progesterone ELISA Kit (Colorimetric)
High sensitivity and specificity for detection of bovine progesterone.
  spinner