EGF ELISA Kits
Invitrogen™ Mouse EGF ELISA Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Assay mouse EGF levels in cell culture supernatants, plasma, serum samples.
R&D Systems™ Human EGF R/ErbB1 Quantikine ELISA Kit, R&D Systems™
a 4.5 hour solid phase ELISA designed to measure EGF R in cell culture supernates, serum, plasma, and human milk.
Invitrogen™ Monkey EGF ELISA Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Assay monkey EGF levels in cell culture supernatants, plasma, serum samples.
R&D Systems™ Human HB-EGF Quantikine ELISA Kit
For the quantitative determination of human Heparin-Binding EGF-like Growth Factor (HB-EGF) concentrations in cell culture supernates, serum, plasma, saliva, urine, and human milk. Human HB-EGF Immunoassay employs the quantitative sandwich enzyme immunoassay technique.
R&D Systems™ Human EGF DuoSet ELISA Kit
Optimized capture and detection antibody pairings with recommended concentrations save lengthy development time
Invitrogen™ Rat EGF ELISA Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Assay rat EGF levels in cell culture supernatants, plasma, serum samples.
  spinner