ELISA Equipment, Labware and Accessories
R&D Systems™ DuoSet Ancillary Reagent Kit 2 Available on GSA/VA Contract for Federal Government customers only.
Sufficient for at least five 96-well plates
Thermo Scientific™ Pierce™ 96-Well Microplates Available on GSA/VA Contract for Federal Government customers only.
Bind antibodies with high capacity using these specially formulated polystyrene plates for ELISA and other immunoassays.
Thermo Scientific™ Pierce™ Stop Solution for TMB Available on GSA/VA Contract for Federal Government customers only.
Stop TMB-based ELISA detection with this 0.16M sulfuric acid for use with the ELISA substrate 3,3',5,5' - tetramethylbenzidine (TMB).
Thermo Scientific™ Microplate Sealing Tape
Clear, self-sticking sealing tape for standard microplates.
Thermo Scientific™ Pierce™ Reagent Reservoirs
Rapidly dispense reagents with multi-channel pipettors using these sterile, sturdy and disposable reservoirs molded from high-impact polystyrene.
Thermo Scientific™ Pierce™ 8-Well Strip Microplates
Pierce 8-Well Strip Plates are optically clear, polystyrene plates that have a standard 96-well microplate footprint but also allow individual 8-well strips (i.e., plate columns) to be removed from the rigid frame.
R&D Systems™ Quantikine Immunoassay Control Group 1 Available on GSA/VA Contract for Federal Government customers only.
Control for Human G-CSF, GM-CSF, IL-1 alpha, IL-1 beta, IL-2, IL-4, IL-6, IL-10, IL-12p70, IFN-gamma, MCP-1, TGF-beta1, TNF-alpha, and VEGF Quantkine ELISAs
Thermo Scientific™ Pierce™ 1.1mL Microtubes and Racked Systems
Pre-racked 1.1mL-tall microtubes boxed to match a standard 96-well microplate footprint for multichannel pipetting or automated workstations.
Thermo Scientific™ Pierce™ Immunodiffusion Plates
Identify and characterize antibody-antigen binding interactions with these standard Ouchterlony gel plates with patterns of six wells around one well.
R&D Systems™ ELISA Plate-coating Buffer Available on GSA/VA Contract for Federal Government customers only.
For use as a plate-coating buffer in ELISA development.
R&D Systems™ Reagent Additive 1
Heat-inactivated Normal Goat Serum, Diluted
MP Biomedicals™ Immunidiffusion (RID) Plates
Used for double and single radial immunodiffusion techniques
Thermo Scientific™ Pierce™ DNA Coating Solution
Efficiently immobilize DNA to microplate wells and microsample tubes with this DNAse-free coating solution.
R&D Systems™ Lysis Buffer 16
R&D Systems™ Lysis Buffer 16 is 21mL of a denaturing buffered solution with preservatives.
Invitrogen™ eBioscience™ InstantOne ELISA Wash Buffer, 10x Non-distribution product as customer accommodation.
Developed for use with the InstantOne ELISA Cell Signaling assays. The InstantOne ELISA Wash Buffer is supplied as a 10x concentrate and is used for washing the InstantOne ELISA cell signaling assay microplate. It is a simple mix of buffers, salts and mild detergent.
Invitrogen™ eBioscience™ InstantOne ELISA Enhancer Solution Non-distribution product as customer accommodation.
Added to the 5x Cell Lysis Buffer (Cat. No. IOCLB1) to yield the Cell Lysis Mix solution that is applied to cultured cells or tissues prior to running the assays.
R&D Systems™ Quantikine Immunoassay Control Group 2
Control for Human IL-1 RII, IL-2 R alpha, IL-6 R alpha, TNF RI, TNF RII, VEGF R1, VEGF R2 Quantikine ELISAs
R&D Systems™ Reagent Diluent Concentrate 3
5X Concentrated 25% Tween 20 solution in PBS for DuoSets
EUROIMMUN Salt for PBS Buffer and Tween 20
These materials, used to create a PBS buffer for use on some Dynex platforms, help to control assay drift, and work to provide the best sensitivity and specificity in results
  spinner