IFN gamma ELISA Kits
R&D Systems™ Human IFN-gamma Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 4.5 hour solid phase ELISA designed to measure IFN-gamma levels in cell culture supernates, serum, and plasma.
R&D Systems™ Mouse IFN-gamma Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 4.5 hour solid phase ELISA designed to measure mouse IFN-gamma in cell culture supernates and mouse serum.
Invitrogen™ Human IFN gamma ELISA Kits Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Measure levels of human interferon gamma (IFNg) in human serum, plasma (EDTA, heparin, sodium citrate), and culture supernatant samples using 96-well plates.
R&D Systems™ Porcine IFN-gamma Quantikine ELISA Kit, R&D Systems™
a 4 hour solid-phase ELISA designed to measure porcine IFN-gamma in cell culture supernates, serum, and plasma.
Invitrogen™ Rat IFN gamma ELISA Kits Non-distribution product as customer accommodation.
Measure levels of rat interferon gamma (IFNg) in rat serum, plasma (EDTA, heparin, sodium citrate), and culture supernatant samples using 96-well plates.
R&D Systems™ Rat IFN-gamma Quantikine ELISA Kit, R&D Systems™
a 4.5 hour solid phase ELISA designed to measure rat IFN-gamma levels in cell culture supernates and serum.
Canine IFN-gamma ELISA Kit, Thermo Scientific™ Pierce™ Non-distribution product as customer accommodation.
Assay canine IFN-gamma levels in cell culture supernatants, plasma, serum samples
Abnova™ IFNG (Human), Biotin, ELISA Kit
IFNG (Human) ELISA Kit is intended for the quantitative measurement of human IFNG.
  spinner