IL-1 alpha ELISA Kits
R&D Systems™ Human IL-1 alpha/IL-1F1 Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 3.5 or 4.5 hour solid phase ELISA designed to measure IL-1 alpha in cell culture supernates, serum, and plasma.
R&D Systems™ Human IL-1ra/IL-1F3 Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 4.5 hour solid phase ELISA designed to measure IL-1ra in cell culture supernates, serum, and plasma.
R&D Systems™ Human IL-1 RII Quantikine ELISA Kit, R&D Systems™
a 3.5 hour solid phase ELISA designed to measure IL-1 sRII in serum, plasma, and cell culture supernates.
R&D Systems™ Rat IL-1 alpha/IL-1F1 Quantikine ELISA Kit, R&D Systems™
a 4.5 hour solid phase ELISA designed to measure rat IL-1 alpha levels in cell culture supernates, serum, and plasma.
R&D Systems™ Mouse IL-1 alpha/IL-1F1 Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 4.5 hour solid phase ELISA designed to measure mouse IL-1 alpha levels in cell culture supernates and serum.
Invitrogen™ Novex IL-1RA Human ELISA Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Designed to quantify protein levels of human interleukin-1 receptor antagonist (IL-1RA) in biological samples
  spinner