IL-10 ELISA Kits
R&D Systems™ Human IL-10 Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 3.5 or 4.5 hour solid phase ELISA designed to measure IL-10 in cell culture supernates, serum, and plasma.
Invitrogen™ Novex EASIA IL-10 Human ELISA Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Designed to quantify protein levels of human IL-10 in serum, plasma, supernatant, and other biological fluids
R&D Systems™ Porcine IL-10 Quantikine ELISA Kit, R&D Systems™
a 4.5 hour solid phase ELISA designed to measure porcine IL-10 levels in cell culture supernates, porcine serum, and EDTA plasma.
R&D Systems™ Mouse IL-10 Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 4.5 hour solid phase ELISA designed to measure mouse IL-10 in cell culture supernates, serum, and plasma.
Invitrogen™ Novex IL-10 Swine ELISA Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
A lymphokine involved in immune response
R&D Systems™ Human IL-10 Quantikine HS ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 5.5 hour solid phase ELISA designed to measure IL-10 in serum and plasma.
Invitrogen™ Human IL-10 ELISA Kits Non-distribution product as customer accommodation.
Measure levels of human interleukin-10 in human serum, plasma (EDTA, heparin, sodium citrate), urine and culture supernatant samples using 96-well plates.
R&D Systems™ Rat IL-10 Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 4.5 hour solid phase ELISA designed to measure rat IL-10 levels in cell culture supernates, serum, and plasma.
Canine IL-10 ELISA Kit, Thermo Scientific™ Pierce™ Non-distribution product as customer accommodation.
Assay canine IL-10 levels in cell culture supernatants, plasma, serum samples
Invitrogen™ Novex IL-10 Human ELISA Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Alymphokine involved in immune response
Invitrogen™ Equine IL-10 ELISA Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Assay equine IL-10 levels in cell culture supernatants, plasma, serum samples
R&D Systems™ Canine IL-10 Quantikine ELISA Kit, R&D Systems™
Sufficient for at least one 96-well plates
Abnova™ IL10 (Human), Biotin, ELISA Kit
IL10 (Human) ELISA Kit is intended for the quantitative measurement of human IL10.
  spinner