IL-16 ELISA Kits
R&D Systems™ Human IL-16 Quantikine ELISA Kit, R&D Systems™
a 4.5 hour solid phase ELISA designed to measure human IL-16 in cell culture supernates, serum, and plasma.
Thermo Scientific™ Pierce™ Human IL-16 ELISA Kits
Measure levels of human interleukin-16 in human serum, plasma (EDTA, heparin, sodium citrate) and culture supernatant samples using 96-well plates. Thermo Scientific™ Interleukin 16 Recombinant protein SIL16 is validated for ELISA and Western Blot.
  spinner