IL-2 ELISA Kits
R&D Systems™ Human IL-2 Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
A 4.5 hour solid phase ELISA designed to measure IL-2 levels in cell culture supernates, serum, and plasma. It contains E. coli-expressed recombinant human IL-2 and antibodies raised against recombinant human IL-2.
Invitrogen™ Human IL-2 ELISA Kits Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Interleukin 2 Recombinant protein SIL2 from Thermo Scientific™ specifically detects Interleukin 2 in samples, and it is validated for ELISA and Western Blot
R&D Systems™ Mouse IL-2 Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 4.5 hour solid phase ELISA designed to measure mouse IL-2 levels in cell culture supernates and serum.
R&D Systems™ Rabbit IL-2 DuoSet, 15 Plate
Sufficient for at least 15 96-well plates
Invitrogen™ Bovine IL-2 ELISA Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Assay bovine Bovine IL-2 levels in cell culture supernatants, plasma, serum samples
R&D Systems™ Rat IL-2 Quantikine ELISA Kit, R&D Systems™
a 4.5 hour solid phase ELISA designed to measure rat IL-2 levels in cell culture supernates, serum, and plasma.
Invitrogen™ Equine IL-2 ELISA Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Assay equine IL-2 levels in cell culture supernatants, plasma, serum samples
R&D Systems™ Human IL-2 QuantiGlo ELISA Kit, R&D Systems™
a 5.5 hour solid phase ELISA designed to measure IL-2 levels in cell culture supernates, serum, and plasma.
  spinner