IL-2 Receptor ELISA Kits
R&D Systems™ Human CD25/IL-2 R alpha Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
A 3.5 hour solid phase ELISA designed to measure IL-2 sR alpha in cell culture supernates, serum, and plasma. It contains NS0-expressed recombinant human IL-2 sR alpha and antibodies raised against the recombinant factor.
Invitrogen™ Human IL-2 Receptor ELISA Kits Non-distribution product as customer accommodation.
Measure levels of human soluble interleukin-2 receptor (sIL-2R, sCD25) in human serum, plasma and culture supernatant samples using 96-well plates.
  spinner