In-Cell ELISA Kits for Specific Targets
R&D Systems™ Human IGF-I Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 3.5 hour solid-phase ELISA designed to measure human IGF-I in cell culture supernates, serum, and plasma.
R&D Systems™ Human sCD14 Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
A 4.5-hour solid-phase ELISA designed to measure human soluble CD14 in cell culture supernants, serum and plasma.
R&D Systems™ Human IL-15 Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 4.25 or 4.5 hour solid phase ELISA designed to measure human IL-15 in cell culture supernates, serum, and plasma.
R&D Systems™ Endothelin-1 Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 4.5 hour solid phase ELISA designed to measure ET-1 in cell culture supernates, serum, plasma, and urine.
R&D Systems™ Human CXCL4/PF4 Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 4.5 hour solid-phase ELISA designed to measure human PF4 in cell culture supernates, serum, and platelet-poor plasma.
R&D Systems™ Mouse Proprotein Convertase 9/PCSK9 Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 4.5 hour solid-phase ELISA designed to measure mouse PCSK9 in cell culture supernates, cell lysates, serum, and plasma.
R&D Systems™ Human Osteopontin (OPN) Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 4.5 hour solid-phase ELISA designed to measure human Osteopontin in cell culture supernates, plasma, urine, and human milk.
R&D Systems™ Human CD163 Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
Sufficient for at least one 96-well plates
R&D Systems™ Human sTNF RI/TNFRSF1A Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
A 4.5 hour solid phase ELISA designed to measure sTNF RI in cell culture supernates, serum, plasma, and urine. It contains E. coli-expressed recombinant human sTNF RI, as well as antibodies raised against this polypeptide.
R&D Systems™ Rat TIM-1/KIM-1/HAVCR Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 4.5 hour solid-phase ELISA designed to measure rat TIM-1 in cell culture supernates, tissue homogenates, serum, plasma, and urine.
R&D Systems™ Human CXCL10/IP-10 Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 4.5 hour solid-phase ELISA designed to measure human IP-10 in cell culture supernates, serum, plasma, saliva, and urine.
Invitrogen™ Novex™ Aβ 42 Human Ultrasensitive ELISA Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Quantifies natural and synthetic Hu Aβ42 in tissue culture medium, tissue homogenate, cerebrospinal fluid (CSF) samples
R&D Systems™ Human Cystatin C Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 4.5 hour solid-phase ELISA designed to measure human Cystatin C in cell culture supernates, serum, plasma, saliva, urine, and human milk.
R&D Systems™ Mouse Lipocalin-2/NGAL Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 4.5 hour solid phase ELISA designed to measure mouse Lipocalin-2 in cell culture supernates, mouse serum, platelet-poor plasma, and urine.
R&D Systems™ Human TGF-beta 1 Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
A 4.5 hour solid phase ELISA designed to measure TGF-beta 1 in acid activated cell culture supernates, serum, plasma, and urine. It contains recombinant human TGF-beta 1 expressed by CHO cells
R&D Systems™ Mouse ST2/IL-1 R4 Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 4.5 hour solid phase ELISA designed to measure mouse ST2 in cell culture supernates, serum, and plasma.
R&D Systems™ Human C-Reactive Protein/CRP Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 4.5 hour solid-phase ELISA designed to measure human CRP in cell culture supernates, serum, and plasma.
Invitrogen™ Novex™ Aβ40 Human ELISA Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Quantifies natural and recombinant human Aβ40 (Beta Amyloid 40) selectively in tissue culture medium, tissue homogenate, and cerebrospinal fluid (CSF)
R&D Systems™ Human Erythropoietin Quantikine IVD ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
Designed to measure Epo levels in serum or plasma in less than 4.5 hours (or less than 2.5 hours using the shaker protocol) for the determination of erythropoietin (Epo) concentration in human serum and plasma as an aid in the diagnosis of anemia and polycythemia. For in vitro diagnostic use.
R&D Systems™ Prostaglandin E2 Parameter Assay Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 3.5 hour forward sequential competitive enzyme immunoassay designed to measure PGE2 in cell culture supernates, serum, plasma, and urine
R&D Systems™ Human FGF-21 Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 4.5 hour solid-phase ELISA designed to measure human FGF-21 in cell culture supernates, serum, and plasma.
R&D Systems™ Human/Mouse/Rat Activin A Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 5.0 hour solid-phase ELISA designed to measure Activin A in human, mouse, or rat cell culture supernates, serum, plasma, and saliva.
R&D Systems™ Mouse/Rat/Porcine/Canine TGF-beta 1 Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 4.5 hour solid phase ELISA designed to measure TGF-beta 1 in acid activated cell culture supernates, serum, and plasma.
Invitrogen™ Novex™ NP40 Cell Lysis Buffer Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Suitable for preparation of cell extracts
R&D Systems™ Mouse Erythropoietin Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 4.5 hour solid-phase ELISA designed to measure mouse Epo in cell culture supernates, tissue homogenates, serum, and plasma.
R&D Systems™ Mouse/Rat IL-22 Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 4.5 hour solid-phase ELISA designed to measure mouse and rat IL-22 in cell culture supernates, serum, and plasma.
BD Mouse IFN-γ ELISA Set
For immunoassay protocols
R&D Systems™ Human Resistin Quantikine ELISA Kit, R&D Systems™
a 4.5 hour solid-phase ELISA designed to measure human Resistin in cell culture supernates, serum, and plasma.
  spinner