Kinase ELISA Kits
Invitrogen™ Phospho GSK-3 beta ELISA Kit Non-distribution product as customer accommodation.
Measure phosphorylated GSK-3b ([pSer9] glycogen synthase kinase 3 beta) levels in cell lysates of human, mouse, and rat origin; coated 96-well plates included.
Invitrogen™ Total MEK 1 ELISA Kit Non-distribution product as customer accommodation.
Measure pan MAPK-ERK kinase 1 (MEK1 and pMEK1, phospho-MEK1) levels in cell lysates of human, mouse, and rat origin in coated 96-well plates.
Invitrogen™ Phospho p38 ELISA Kit Non-distribution product as customer accommodation.
Measure phosphorylated p38 ([pThr180/Tyr182] p38 MAP kinase, phospho-p38) levels in cell lysates of human and mouse origin; coated 96-well plates included.
Invitrogen™ Phospho ERK 1/2 ELISA Kit, Thermo Scientific™ Non-distribution product as customer accommodation.
Microplate sandwich ELISA kit for phospho-ERK (pERK12)
Invitrogen™ Phospho MEK 1 ELISA Kit Non-distribution product as customer accommodation.
Measure phosphorylated MEK-1 ([pSer218/Ser222] MAPK-ERK kinase 1) levels in cell lysates of human, mouse, and rat origin; coated 96-well plates included.
Invitrogen™ Phospho JNK 1/2 ELISA Kit Non-distribution product as customer accommodation.
Measure phosphorylated JNK-1 and JNK-2 ([pThr183/Tyr185] c-Jun N-terminal protein kinase) levels in cell lysates of human, mouse, and rat origin; coated 96-well plates included.
  spinner