MCP-1 ELISA Kits
R&D Systems™ Human CCL2/MCP-1 Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 3.5 or 4.5 hour solid phase ELISA designed to measure MCP-1 in cell culture supernates, serum, plasma, and urine.
  spinner