Agglutinins
MP Biomedicals™ Wheat Germ Agglutinin, 5mg
isolated from wheat germ, dissolved in 0.05 M Tris-HCl, 0.15 M NaCl buffer, pH 8.6 and purified by affinity chromatography
MP Biomedicals™ Lotus tetragonolobus Agglutinin
Exhibits a specificity for α-L-fucose.
  spinner