Streptavidin Products
Invitrogen™ DYNAL™ MyOne™ Dynabeads™ Streptavidin C1 Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Exceptional standard for isolation and handling of biotinylated nucleic acids, antibodies, or other biotinylated ligands and targets
Invitrogen™ DYNAL™ MyOne™ Dynabeads™ Streptavidin T1 Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Exceptional standard for isolation and handling of biotinylated nucleic acids, antibodies, or other biotinylated ligands and targets
BD Streptavidin HRP Solution
Streptavidin is a non-glycosylated protein that is prepared chromatographically from the bacterium Streptomyces avidinii. Streptavidin homotetramers have a particularly high, non-covalent binding affinity for biotin.
Thermo Scientific™ Pierce™ Poly-HRP Streptavidin
An advanced polymer of HRP conjugated to streptavidin with a high HRP:streptavidin ratio
BD Streptavidin Particles Plus - DM
For positive selection or depletion of leukocyte subpopulations using the BD IMagnet™
Invitrogen™ DYNAL™ Dynabeads™ Streptavidin Trial Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Economical trial kit lets you test Dynabeads in your application
R&D Systems™ EvenCoat Streptavidin Coated Plates Available on GSA/VA Contract for Federal Government customers only.
used as a solid support for assays that utilize a biotinylated molecule or otherwise require the binding of biotin.
GE Healthcare Streptavidin Mag Sepharose™ Magnetic Bead Available on GSA/VA Contract for Federal Government customers only.
For simple and efficient enrichment of target proteins by immunoprecipitation and purification of biotinylated biomolecules
R&D Systems™ Streptavidin-HRP Available on GSA/VA Contract for Federal Government customers only.
For use in ELISAs, dilute to the working concentration specified on the vial label using 1% BSA/PBS or Reagent Diluent Concentrate 2 (Catalog # DY995) diluted 1:10 in deionized water.
Thermo Scientific™ Pierce™ HRP-Conjugated Streptavidin
Streptavidin from Streptomyces avidinii conjugated to highly purified horseradish peroxidase (HRP)
BD ELISPOT Streptavidin-HRP for ELISPOT
Recommended for use with BD ELISPOT Pairs
BD Horizon Brilliant™ Ultraviolet 496 Strepavidin
Streptavidin is a non-glycosylated protein that is prepared chromatographically from the bacterium Streptomyces avidinii
SouthernBiotech™ Streptavidin Reagents
Offer less nonspecific binding than egg white avidin
Thermo Scientific™ Lab Vision™ Ready-To-Use Streptavidin Peroxidase
Incubate slides for 10 minutes with Thermo Scientific™ Lab Vision™ Streptavidin Peroxidase (Ready-to-Use) reagent for results that will remain stable for 18 months.
R&D Systems™ Streptavidin-Allophycocyanin
Optimized for use with biotin-conjugated monoclonal antibodies
R&D Systems™ Streptavidin-Phycoerythrin
Optimized for use with monoclonal antibody-biotin reagents
BD Horizon Brilliant™ Violet 480 Strepavidin
Streptavidin has very high binding affinity for biotin, which is conjugated to the antibody
BD Pharmingen™ Streptavidin-HRP
Prediluted for convenient use
R&D Systems™ Streptavidin-Alkaline Phosphatase
Intended for use in ELISA and immunoblotting assays
BD AKP Streptavidin
Pre-diluted, ready-to-serve, convenient to use enzyme which provides optimal signal amplification and detection of an antigen
R&D Systems™ NorthernLights Streptavidin, NL637
For use as a secondary developing reagent in immunofluorescence assays with biotinylated primary antibodies
  spinner