Western Blotting Reagents, Membranes, Supplies
Thermo Scientific™ UltraVision™ Quanto Detection System HRP
Deliver higher lab profitability and better staining by selecting the best antibody for each test with the Thermo Scientific™ UltraVision™ Quanto HRP DAB.
Thermo Scientific™ Lab Vision™ MultiVision Polymer Detection System: anti-Mouse-HRP and anti-Rabbit-AP
A simple and robust one-step polymer detection system for visualization of two antigens simultaneously in blue and red.
Thermo Scientific™ SuperSignal™ West Femto Chemiluminescent Substrate Available on GSA/VA Contract for Federal Government customers only.
Ultra-sensitive enhanced chemiluminescent (ECL) HRP substrate for low-femtogram-level detection by Western blot analysis.
Thermo Scientific™ Restore™ Western Blot Stripping Buffer Available on GSA/VA Contract for Federal Government customers only.
Safely and effectively remove primary and secondary antibodies from nitrocellulose and PVDF membranes to allow chemiluminescent Western blots to be reprobed.
MilliporeSigma&trade Immobilon™-P PVDF Transfer Membranes Available on GSA/VA Contract for Federal Government customers only.
Optimized for binding proteins transferred from a variety of gel matrices. MilliporeSigma™ Immobilon™-P PVDF Transfer Membranes have superior handling, staining, and resistance than nitrocellulose. For use with Western blots and protein sequencing. Available in sheets or rolls.
Invitrogen™ Novex™ iBlot™ 2 Nitrocellulose Transfer Stacks Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Used to transfer proteins with the iBlot 2 Gel Transfer Device
Thermo Scientific™ Pierce™ Recombinant Protein L and Conjugates Available on GSA/VA Contract for Federal Government customers only.
Detect mouse and human antibodies containing kappa light chains in immunoassays using this purified, biotinylated or peroxidase-conjugated, recombinant Protein L.
Thermo Scientific™ Pierce™ Streptavidin Poly-HRP Conjugate Available on GSA/VA Contract for Federal Government customers only.
Amplify signal and improve detection of biotinylated antibodies for IHC and other methods using this streptavidin conjugated with HRP polymers.
Thermo Scientific™ SuperSignal™ Western Blot Enhancer
Increase both signal intensity and sensitivity 3- to 10-fold with this membrane treatment reagent and primary antibody diluent.
Thermo Scientific™ Clean-Blot IP Detection Reagents and Kit
Avoid interference from denatured IP antibody fragments in Western blots of immunoprecipitated proteins with these HRP and AP conjugates.
Thermo Scientific™ Owl™ HEP Series Semidry Electroblotting Systems Available on GSA/VA Contract for Federal Government customers only.
Provide faster blots with higher efficiency using Thermo Scientific™ Owl™ HEP Series Semidry Electroblotting Systems.
Thermo Scientific™ Pierce™ Western Blot Signal Enhancer
Greatly enhance the effectiveness of primary antibodies and intensify the final Western blot detection signal with this two-reagent blot conditioning system.
Thermo Scientific™ Pierce™ Streptavidin HRP Conjugate, High Sensitivity Available on GSA/VA Contract for Federal Government customers only.
Amplify signal in Western blot, ELISA or other biotin-based detection methods with this exclusive, high signal-to-noise, peroxidase-conjugated, ready-to-use streptavidin.
Thermo Scientific™ CL-XPosure™ Film (X-Ray Film) Available on GSA/VA Contract for Federal Government customers only.
Capture results of ECL-based Western blots and other chemiluminescent protein or nucleic acid assays using this economically priced, clear-blue X-ray film.
Thermo Scientific™ Pierce™ Recombinant Protein G and Conjugates
Detect mouse and human antibodies, especially IgG isotypes, in immunoassays with this purified, biotinylated or peroxidase-conjugated recombinant Protein G.
MilliporeSigma&trade Immobilon™-FL PVDF Transfer Membranes Available on GSA/VA Contract for Federal Government customers only.
Designed specifically for Western blots and protein sequencing. MilliporeSigma™ Immobilon™-FL PVDF Transfer Membranes are plain surfaced with 0.45μm pores and have lower background fluorescence than other membranes. Available in 10cm and 20cm squares and a 3.75m roll.
Thermo Scientific™ Pierce™ Maleimide-Activated HRP Conjugation Kit
Activated horseradish peroxidase (HRP) enzyme and kit to label proteins, peptides or other ligands via sulfhydryl groups (reduced cysteines) to make HRP conjugates.
Thermo Scientific™ Blocker™ Casein Blocking Buffers Available on GSA/VA Contract for Federal Government customers only.
Block membranes, plates and other samples with these ready-to-use PBS or TBS solutions of purified casein for Western blot, ELISA and other detection methods.
Invitrogen™ Molecular Probes™ CellROX Green Reagent Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Novel fluorogenic probe for measuring oxidative stress in live cells
Thermo Scientific™ Restore™ Plus Western Blot Stripping Buffer Available on GSA/VA Contract for Federal Government customers only.
Remove high-affinity antibodies from membranes so they can be reprobed and detected with chemiluminescent substrates.
Thermo Scientific™ Lab Vision™ UltraVision™ LP Detection System: HRP Polymer (Ready-To-Use)
Employ a senstive detection system based on biotin-free, polymer technology.
MilliporeSigma&trade Luminata Forte Western HRP substrate, 100mL Available on GSA/VA Contract for Federal Government customers only.
Premixed, ready to use chemiluminescent HRP detection reagent
Invitrogen™ Novex™ iBlot™ 2 PVDF Transfer Stacks Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Consumable stacks with integrated PVDF transfer membranes for dry blotting of proteins
Thermo Scientific™ SuperSignal™ West Pico PLUS Chemiluminescent Substrate Available on GSA/VA Contract for Federal Government customers only.
An enhanced chemiluminescent (ECL) horseradish peroxidase (HRP) substrate that enables picogram- to high femtogram-level protein detection by western blot analysis
Thermo Scientific™ Pierce™ PVDF Membranes Available on GSA/VA Contract for Federal Government customers only.
Enable high protein and nucleic acid binding capacity with sheets and rolls of high-quality polyvinylidene difluoride (PVDF) membrane.
Invitrogen™ iBlot™ 2 Gel Horizontal Transfer Device Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Performs Western blotting transfer simply, efficiently, and reliably, within seven minutes and without the need for liquid buffers
NeutrAvidin Protein, Invitrogen™ Available on GSA/VA Contract for Federal Government customers only.
NeutrAvidin Protein, 31000, from Invitrogen™, 31000, from Invitrogen™ Species Reactivity: ; Applications: ELISA, Immunohistochemistry, In Situ Hybridization (ISH), Western Blot.
Invitrogen™ Molecular Probes™ CellROX™ Deep Red Reagent Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Optimized probe for detection of oxidative stress in cells
  spinner