Blocking Buffers for Protein Methods
Thermo Scientific™ Blocker™ Casein Blocking Buffers
Block membranes, plates and other samples with these ready-to-use PBS or TBS solutions of purified casein for Western blot, ELISA and other detection methods.
Thermo Scientific™ Blocker™ BSA Blocking Buffers
Block membranes, plates and other samples with these ready-to-use 10X PBS or TBS solutions of bovine serum albumin for Western blot, ELISA and other detection methods.
Thermo Scientific™ Blocker™ BLOTTO Blocking Buffer
Block membranes, plates and other samples with this ready-to-use 5% solution of nonfat powdered milk in TBS for Western blot, ELISA and other detection methods.
Thermo Scientific™ Pierce™ Protein-Free Blocking Buffers Available on GSA/VA Contract for Federal Government customers only.
Block membranes, plates and other samples with these ready-to-use PBS and TBS formulations of a non-protein compound to yield extremely low background.
Thermo Scientific™ Pierce™ Clear Milk Blocking Buffer
Thermo Scientific™ Pierce Clear Milk Blocking Buffer is a long shelf-life, high performance replacement for homemade milk blocking buffers in Western blotting applications.
Thermo Scientific™ SEA BLOCK Blocking Buffer
Thermo Scientific™ SEA BLOCK Blocking Buffer is steelhead salmon serum in PBS that is especially useful as a blocking agent in IHC and other detection methods involving mammalian samples
Thermo Scientific™ Pierce™ Fast Blocking and Wash Buffers
Optimize Western blotting systems by decreasing both background signal and incubation time compared to traditional blocking agents with this ready-to-use blocker solution.
MilliporeSigma™ Calbiochem™ OmniPur™ Nonfat Powdered Milk
Serves as blocking agent for Western Blot analysis.
Invitrogen™ eBioscience™ IHC/ICC Blocking Buffer, High Protein Non-distribution product as customer accommodation.
Designed for use in immunohistochemistry and immunocytochemistry protocols which require blocking of non-specific binding sites. IHC/ICC Blocking Buffer, High Protein is recommended for tissues and antibodies with high background staining, nuclear antigens, and FFPE tissue.
Invitrogen™ eBioscience™ IHC /ICC Blocking Buffer, Low Protein Non-distribution product as customer accommodation.
Designed for use in immunohistochemistry and immunocytochemistry protocols which require blocking of non-specific binding sites. IHC /ICC Blocking Buffer, Low Protein is recommended for tissues and antibodies with average levels of background staining.
Novex™ Membrane Blocking Solution Non-distribution product as customer accommodation.
Membrane Blocking Solution
MilliporeSigma™ Novagen™ 5% Alkali - Soluble Casein
Used as blocking reagent for Western blot applications.
FCS Blocking Buffer in TBS
For blocking excess solid surface in ELISA
MilliporeSigma™ Calbiochem™ BLOT-QuickBlocker™ Reagent
Prevents nonspecific proteins from binding to blots.
  spinner