Western Blotting Substrates
Thermo Scientific™ SuperSignal™ West Femto Chemiluminescent Substrate Available on GSA/VA Contract for Federal Government customers only.
Ultra-sensitive enhanced chemiluminescent (ECL) HRP substrate for low-femtogram-level detection by Western blot analysis.
Thermo Scientific™ SuperSignal™ West Dura Chemiluminescent Substrate Available on GSA/VA Contract for Federal Government customers only.
Luminol-based enhanced chemiluminescence (ECL) HRP substrate with very stable light output for femtogram-level detection with CCD-based imagers.
Thermo Scientific™ Pierce™ ECL Western Blotting Substrate
Value-priced, entry-level peroxidase substrate for enhanced chemiluminescence (ECL) that directly replaces costlier products without the need to re-optimize conditions.
GE Healthcare Amersham™ ECL™ Prime Western Blotting Detection Reagent Available on GSA/VA Contract for Federal Government customers only.
Enables the use of highly diluted primary and secondary antibodies without reducing sensitivity.  GE Healthcare Lifescience™ Amersham™ ECL™ Prime Western Blotting Detection Reagent produces high signal intensity and sensitivity.
Thermo Scientific™ Pierce™ ECL 2 Western Blotting Substrate Available on GSA/VA Contract for Federal Government customers only.
Recommended HRP substrate for fluorescence imagers
MilliporeSigma™ Luminata™ Western HRP Chemiluminescence Substrates Available on GSA/VA Contract for Federal Government customers only.
Premixed, ready-to-use chemiluminescent reagents for detection of HRP-based Westerns
Thermo Scientific™ Pierce™ NBT/BCIP Substrate Solutions
Powders and ready-made nitro-blue tetrazolium and 5-bromo-4-chloro-3'-indolyphosphate solutions for chromogenic detection of alkaline phosphatase probes.
GE Healthcare Amersham™ ECL™ Western Blotting Detection Reagents Available on GSA/VA Contract for Federal Government customers only.
Use for all routine Western blotting. GE Healthcare Life Sciences™ Amersham™ ECL™ Western Blotting Detection Reagents are the world’s first commercially available chemiluminescent detection reagents and still one of the most widely used.
Thermo Scientific™ 1-Step™ Ultra TMB-Blotting Solution
Thermo Scientific™ 1-Step Ultra TMB-Blotting Solution is an enhanced single-component horseradish peroxidase (HRP) substrate for Western blotting and immunohistochemistry.
Thermo Scientific™ Pierce™ DAB Substrate Kit
Chromogenic horseradish peroxidase (HRP) substrate for Western blot and tissue staining methods.
GE Healthcare Amersham™ ECL™ Prime Western Blotting Detection Reagent PROMO
Enables the use of highly diluted primary and secondary antibodies without reducing sensitivity.  GE Healthcare Amersham™ ECL™ Prime Western Blotting Detection Reagent produces high signal intensity and sensitivity.
Thermo Scientific™ 1-Step™ TMB-Blotting Substrate Solution
Ready-to-use blue TMB peroxidase (HRP) substrate specially formulated for chromogenic detection in Western blot and IHC experiments.
Thermo Scientific™ Pierce™ CN/DAB Substrate Kit
Combines chloronaphthol- and diaminobenzidine-based HRP detection for Western blot and tissue staining methods.
MilliporeSigma™ BlØk™ Noise Canceling Reagents Available on GSA/VA Contract for Federal Government customers only.
Minimize background in Western blots, increasing sensitivity for superior detection of even low-abundance proteins
Thermo Scientific™ Pierce™ Chloronaphthol (CN) Substrates
Powders, tablets and ready-made 4-chloro-1-naphthol solutions for chromogenic HRP detection in Western blot and tissue staining methods.
  spinner