Bottletop Filters
Thermo Scientific™ Nalgene™ Rapid-Flow™ Sterile Disposable Bottle Top Filters with PES Membrane Available on GSA/VA Contract for Federal Government customers only.
Perform sterile filtration of cell culture media and serum with Thermo Scientific™ Nalgene™ Rapid-Flow™ Sterile Disposable Bottle Top Filters with PES Membranes.
Corning™ Disposable Sterile Bottles Available on GSA/VA Contract for Federal Government customers only.
Use as replacement receivers for Corning Disposable Sterile Filter Systems or as storage bottles. Corning™ Disposable Sterile Bottles are easy to grip and ideal for storage of media, buffers and other aqueous solutions.
MilliporeSigma™ Steritop Threaded Bottle Top Filter
The Stericup™ vacuum filtration system is the most efficient single-use, disposable process for sterile media preparation. It combines a Steritop™ filter unit with a receiver flask for processing and storing volumes from 150 mL to 1,000 mL in a single, ready-to-use format.
MilliporeSigma™ Stericup Quick Release-GP Sterile Vacuum Filtration System
The Stericup™ vacuum filtration system is the most efficient single-use, disposable process for sterile media preparation. It combines a Steritop™ filter unit with a receiver flask for processing and storing volumes from 150 mL to 1,000 mL in a single, ready-to-use format.
Corning™ Disposable Vacuum Filter/Storage Systems Available on GSA/VA Contract for Federal Government customers only.
Features printed funnels that identify membrane type and product number for easy product product identification. Corning™ Disposable Vacuum Filter/Storage Systems provide a complete solution for your filtering needs. The system consists of a polystyrene filter funnel joined by an adapter ring to a removable polystyrene storage bottle with a separate sterile polyethylene cap.
MilliporeSigma™ Millipak™ Disposable Filter Units Available on GSA/VA Contract for Federal Government customers only.
Removes particles and microorganisms from liquids and gases with pump-driven filter. Merck Millipore™ Millipak™ Disposable Filter Units provide sterile filtration of parenterals, diagnostics, buffers, media, and biologicals.
Corning™ Centrifuge Tube Top Vacuum Filters Available on GSA/VA Contract for Federal Government customers only.
Filters directly into centrifuge tube minimizing unnecessary transfers. Corning™ Centrifuge Tube Top Filters offer a viewing slot in base that lets you measure volumes as low as 5mL.
Corning™ Disposable Sterile Bottle-Top Filters with 0.22μm Membrane Available on GSA/VA Contract for Federal Government customers only.
Designed for use on glass or polystyrene media bottles with 33 and 45mm neck sizes. Corning™ Disposable Sterile Bottle-Top Filters are ideal for filtration of tissue culture media and components, biological fluids, other solutions.
Thermo Scientific™ Nalgene™ Rapid-Flow™ Sterile Disposable Bottle Top Filters with SFCA Membrane Available on GSA/VA Contract for Federal Government customers only.
Lower the number of extractables in your bottle-top filters by switching from standard CA membranes to sterile, disposable bottle top filters with SFCA Membrane.
MilliporeSigma™ Millex™-GP50 Sterile Filter Unit Available on GSA/VA Contract for Federal Government customers only.
Fast filtration with low protein binding
Thermo Scientific™ Disposable Filter Columns Available on GSA/VA Contract for Federal Government customers only.
Disposable polypropylene columns effectively filter out interfering materials from blood serum or urine
MilliporeSigma™ Steritop™ Sterile Vacuum Bottle-Top Filters
Filter and sterilize faster with a large 7.3cm diameter filter. MilliporeSigma™; Steritop™ Sterile Vacuum Bottle-Top Filters are sterile, threaded bottle-top filters with 0.22 micron pore size for vacuum filtration and sterilization of tissue culture media, protein and DNA solutions, and virus suspensions.
MilliporeSigma™ Sterivex Sterile Pressure-Driven Devices Available on GSA/VA Contract for Federal Government customers only.
Minimize contamination risk to filtered solution by direct-dispensing design. MilliporeSigma™ Sterivex Sterile Pressure-Driven Devices are ready-to-use units that filter solutions with volumes of 100 to 2000mL. Available in a variety of fittings, pore sizes and membrane types.
MilliporeSigma™ Stericap™ PLUS Sterile Bottle-Top Filter Device
For ultrafast, aseptic sterilization of 5 to 10L using vacuum filtration
Fisherbrand™ Disposable PES Bottle Top Filters
Increase sample protection with Fisherbrand Sterile Disposable Bottle Top Filters with PES Membranes. The polyethersulfone (PES) membranes have low extractables and low protein binding for use in sterile filtration and clarification of cell culture media and serum.
MilliporeSigma™ Millistak+™ HC Pod Disposable Depth Filter System
Combine two distinct technologies to enhance filter capacity and retention
Thermo Scientific™ Nalgene™ Large-Volume FastCap™ Bottle Top Filter with 0.2μm PES Membrane Available on GSA/VA Contract for Federal Government customers only.
Filter up to 5L of fluid with this large-volume bottle top filter with a 0.2μm PES membrane. Place on a bottle mouth, apply vacuum and start the filtering process.
MilliporeSigma™ Millex™ LCR Multi-Layer Filter Available on GSA/VA Contract for Federal Government customers only.
Hydrophilic PTFE filters for manual or automated use
Corning™ Disposable Sterile Bottle-Top Filters with 0.45μm Membrane Available on GSA/VA Contract for Federal Government customers only.
Designed for use on glass or polystyrene media bottles with 33 and 45mm neck sizes. Corning™ Disposable Sterile Bottle-Top Filters are ideal for filtration of tissue culture media and components, biological fluids, other solutions.
Spectrum™ MediaKap™ and MediaKap Plus HF Filters
0.2μm DynaFibre™ mixed cellulose ester filter provides more surface area for faster flow than disc filters with less fouling
MilliporeSigma™ IC Millex Nonsterile LH Filter Units Available on GSA/VA Contract for Federal Government customers only.
Low-extractable filter specifically designed for ion chromatography
MilliporeSigma™ Stericap PLUS, 1L Receiver Flask
Used for storage of sterilized or clarified fluids.
DWK Life Sciences Wheaton™ 25mm Filtration Assemblies and Accessories Available on GSA/VA Contract for Federal Government customers only.
Choose from a wide range of filtration assemblies and accessories to suit your application needs
DWK Life Sciences Wheaton™ 47mm Filtration Assembly Available on GSA/VA Contract for Federal Government customers only.
Designed to handle up to 1000mL of liquid for analysis of particulate or microbiological contamination
MilliporeSigma™ Steritop™-VP Bottletop Filter Unit, 10μm
Provides a large filter area for rapid sterile filtration of aqueous fluids into wide-mouth bottles (45mm). MilliporeSigma™ Steritop™-VP Bottletop Filter Unit, 10μm features recessed bottle threads to prevent contamination from filter contact with work surfaces and a stable, no tip–easy grip design.
DWK Life Sciences Wheaton™ 47mm Filtration Assembly and Accessories Available on GSA/VA Contract for Federal Government customers only.
Designed for filtering viscous or proteinaceous solutions to produce ultraclean filtrate. Wheaton™ 47mm Filtration Assembly and Accessories are manufactured from the highest quality materials.
DWK Life Sciences™ Wheaton™ 47mm Filtration Assembly with Standard Taper Joint Connection and Fritted Glass Support Available on GSA/VA Contract for Federal Government customers only.
Recommended for routine filtration of organic solvents, corrosive liquids and the removal of particulates from HPLC solvents. Wheaton™ 47mm Filtration Assembly with Standard Taper Joint Connection and Fritted Glass Support is manufactured from low extractable borosilicate glass.
MilliporeSigma™ Millex™ HPF Filter Units: Pack of 50
Greater throughput for easier filtration
  spinner