Filter Units
Thermo Scientific™ Nalgene™ Rapid-Flow™ Sterile Disposable Filter Units with PES Membrane Available on GSA/VA Contract for Federal Government customers only.
Increase filtration efficiency. PES membranes and exclusive Rapid-Flow design provide fast flow rates and high throughput while defending against cell culture contamination.
MilliporeSigma™ Steriflip™ Sterile Disposable Vacuum Filter Units
Performance and convenience all in one.  Merck Millipore™ Steriflip™ Sterile Disposable Vacuum Filter Units uses vacuum to collect filtrate.
MilliporeSigma™ Stericup Quick Release-GP Sterile Vacuum Filtration System
The Stericup™ vacuum filtration system is the most efficient single-use, disposable process for sterile media preparation. It combines a Steritop™ filter unit with a receiver flask for processing and storing volumes from 150 mL to 1,000 mL in a single, ready-to-use format.
Corning™ Disposable Vacuum Filter/Storage Systems Available on GSA/VA Contract for Federal Government customers only.
Features printed funnels that identify membrane type and product number for easy product product identification. Corning™ Disposable Vacuum Filter/Storage Systems provide a complete solution for your filtering needs. The system consists of a polystyrene filter funnel joined by an adapter ring to a removable polystyrene storage bottle with a separate sterile polyethylene cap.
Thermo Scientific™ Nalgene™ Rapid-Flow™ Sterile Disposable Filter Units with CN Membrane Available on GSA/VA Contract for Federal Government customers only.
Perform sterile filtration, prefiltration or clarification of buffers and other aqueous solutions when protein binding is not a concern with these filter units.
Thermo Scientific™ Nalgene™ Rapid-Flow™ Sterile Disposable Filter Units with SFCA Membrane Available on GSA/VA Contract for Federal Government customers only.
Perform vacuum filtration and single-use biological work requiring sterile filtration, prefiltration or clarification of aqueous solutions in the lab or field.
MilliporeSigma™ Stericup™ Sterile Vacuum Filter Units
Designed for sterilizing tissue culture media, protein and DNA solutions. MilliporeSigma™ Stericup™ Sterile Vacuum Filter Units have a no-tip design for improved stability during filtration. Steritop bottle top filter units use either a PES or PVDF filter. Available in varied pore sizes and volumes.
Fisherbrand™ Disposable PES Filter Units
Increase cell culture sample protection with Fisherbrand sterile disposable vacuum filter units with PES membranes. The polyethersulfone (PES) membranes have low extractables and low protein binding for use in sterile filtration and clarification of cell culture media and serum.
Thermo Scientific™ Nalgene™ Rapid-Flow™ Sterile Disposable Filter Units with Nylon Membrane Available on GSA/VA Contract for Federal Government customers only.
Filter solutions where very low extractables are desired and solutions containing weak solvents with these disposable filter units with nylon membrane.
MilliporeSigma™ Millex-HP Filter Unit Available on GSA/VA Contract for Federal Government customers only.
A 33mm diameter sterile syringe filter with a 0.45μm pore size hydrophilic Polyethersulfone (PES) membrane.
MilliporeSigma™ Millex-MP Filter Unit Available on GSA/VA Contract for Federal Government customers only.
A 25mm diameter sterile syringe filter with a 0.22μm pore size Polyethersulfone membrane. Comes in a pack of 50.
MilliporeSigma™ Nylon-Net Steriflip™ Vacuum Filter Unit
Sterile, disposable filter unit for cell isolation
MilliporeSigma™ Millex-FG Filter Unit
A 25mm diameter sterile syringe filter with a 0.2μm pore size hydrophobic PTFE membrane. Comes in a pack of 50.
Thermo Scientific™ BioLite Filter Units
Safeguard your cells and your budget. Choose economical filter units with aPES membranes for performing general filtration procedures for sterile filtration of your cell culture media.
MilliporeSigma™ Millex-LG Filter Unit Available on GSA/VA Contract for Federal Government customers only.
A 13mm diameter sterile syringe filter with a 0.2μm pore size hydrophilic PTFE membrane. Comes in a pack of 100.
GE Healthcare Whatman™ Glass Vacuum Filtration Devices
Equipment and accessories for vacuum filtration
Sartorius™ Syringe Filter Holders Available on GSA/VA Contract for Federal Government customers only.
Achieve leak-free filtration up to 7bar. Sartorius™ In-Line Filter Holders are inexpensive, reusable holders made of clear polycarbonate with silicone gaskets. Auclavable units are available for 25mm and 13mm syringe filters.
Sartorius™ Biosart™ 100 Monitors
Designed specifically for microbiological testing of pharmaceuticals, food, beverages, water and other liquids. Sartorius™ Biosart™ 100 Monitors are sterile disposables with an incorporated membrane filter and cellulose pad are ready to use. The printed grid lines make the counting easier, especially for higher bacteria counts and for micro-colonies, but does not influence the growth.
Sartorius™ Sartolab™ P20 Plus Filter Systems
Filter tissue culture solutions quickly, economically and reliably. Sartorius™ Sartolab™ P20 Plus Filter Systems are sterile, ready-to-use units. A large filtration area and automatic de-aeration ensures high flow rates. Available with PES or SFCA membrane, different connectors or with a prefilter to process up to 10L.
Sartorius™ Sartolab™ RF/BT Vacuum Filtration Units Available on GSA/VA Contract for Federal Government customers only.
Filter aqueous solutions quickly and easily. Sartorius™ Sartolab™ RF/BT Vacuum Filtration Units are disposable and easy to use filter units. They contain a high quality PES membrane and combine the highest flow-rates and throughput with extremely low protein binding and extractables. Available in a range of volumes.
Sartorius™ Microsart™ Funnels
Ready-to-use, sterile, polypropylene funnels with click-fit closure
Sartorius™ Biosart™ 250 Funnels
Sterile Funnels, ready-to-use
MilliporeSigma™ Millex-GS Filter Unit
25mm syringe filter with a Mixed Cellulose Esters membrane, for non-sterile filtration, has a 0.22μm pore size used in Sterilization of aqueous solutions.
DWK Life Sciences DURAN™ Bottle Top Vacuum Filtration Units
Designed for sterilization or clarification of aqueous cell culture media.
MilliporeSigma™ Millex Samplicity™ Filters, Hydrophilic PTFE
To be used with the Samplicity Filtration System. Powered by vacuum, the system takes minimal manual effort to filter 1 to 8 samples into standard HPLC vials in seconds, with higher recovery than syringe filters provide.
Sartorius™ Polycarbonate Filtration Holders
For the clarification or sterile filtration of up to about 200ml volumes of aqueous solutions
Sartorius™ Equipment and Accessories for Vacuum Filter Holders
Choose the best accessories to support your filtration applications. Sartorius™ Equipment and Accessories for Vacuum Filter Holders include an all-glass vacuum filter holder assembly plus a hose, hand-held pump with gauge, a 3-way valve, a 3-way tap and a stainless steel connector.
  spinner