Qualitative Filter Paper
GE Healthcare Whatman™ Grade 2V Folded Qualitative Filter Papers Available on GSA/VA Contract for Federal Government customers only.
Saves time required to fit circles into conical funnels. Whatman™ Grade 2V Folded Qualitative Filter Papers provide medium flow and nominal particle retention rating of 8µm at a higher filtration speed.
GE Healthcare Whatman™ Qualitative Filter Paper: Grade 1 Circles
Covers a wide range of laboratory applications and is frequently used for clarifying liquids. Whatman™ Qualitative Filter Paper: Grade 1 Circles are widely used standard grade filter papers for routine applications with medium retention and flow rates.
GE Healthcare Whatman™ Quartz Air Sampling Filters, Grade QM-A, Circle Available on GSA/VA Contract for Federal Government customers only.
Used in many air sampling filter applications, including sampling of acidic gases, stacks, flues, and aerosols. Whatman™ Quartz Air Sampling Filters are sequentially numbered according to U.S. Environmental Protection Agency (EPA) standards and are suitable for most applications.
Fisherbrand™ Qualitative Grade Plain Filter Paper Circles - P8 Grade
Ideal for clarifying and removing particles of 20μm and larger, and in applications that do not require a low-ash or ashless paper.
GE Healthcare Whatman™ Qualitative Filter Paper: Grade 4 Circles
Filters and retains course particles for monitoring air pollution. Whatman™ Qualitative Filter Paper: Grade 4 Circles are used when high flow rates in air pollution monitoring are required and the collection of fine particles is not critical.
GE Healthcare Whatman™ Qualitative Filter Paper: Grade 2 Circles
Retains and absorbs more than Grade 1 filters, with a corresponding increase in filtration time. Whatman™ Qualitative Filter Paper: Grade 2 Circles are used for monitoring specific contaminants in the atmosphere and in soil testing.
Fisherbrand™ Wet-Strengthened Qualitative Filter Paper Circles
Coarse cellulose fiber papers remove particles >25μm
GE Healthcare Whatman™ Qualitative Filter Paper: Grade 1 Sheets
Covers a wide range of laboratory applications and is frequently used for clarifying liquids. Whatman™ Qualitative Filter Paper: Grade 1 Sheets are widely used standard grade filter paper for routine applications with medium retention and flow rates.
GE Healthcare Whatman™ Qualitative Grade Plain Circles and Sheets - Grade 3 Available on GSA/VA Contract for Federal Government customers only.
Used for high precipitate capacity and increased wet strength. Whatman™ Qualitative Grade Plain Circles and Sheets - Grade 3 provide double the thickness of Whatman Grade 1.
Fisherbrand™ Qualitative Grade Plain Filter Paper Circles - P5 Grade
Ideal for clarifying and removing particles of 1μm and larger, and in applications that do not require a low-ash or ashless paper
GE Healthcare Whatman™ Qualitative Filter Paper Circles - Grade 2 for IS Measurement
Specifically developed for measuring the Ignition Strength (IS) of Cigarettes. Whatman™ Qualitative Filter Paper Circles - Grade 2 for IS Measurement are manufactured under strict control; each lot is tested and certified as meeting the requirements of ASTM E2187.
Fisherbrand™ Qualitative Grade Plain Filter Paper Circles and Sheets - P4 Grade
Ideal for clarifying and removing particles of 1μm and larger, and in applications that do not require a low-ash or ashless paper
GE Healthcare Whatman™ Shark Skin Filter Paper Available on GSA/VA Contract for Federal Government customers only.
High wet strength allows use under pressure
Fisherbrand™ Qualitative Grade Plain Filter Paper Sheets - P8 Grade
Ideal for clarifying and removing particles of 1μm and larger, and in applications that do not require a low-ash or ashless paper
Fisherbrand™ Qualitative Grade Plain Filter Paper Sheets - P5 Grade
Ideal for clarifying and removing particles of 1μm and larger, and in applications that do not require a low-ash or ashless paper
Bel-Art™ SP Scienceware™ Filter Paper for Table-Top Buchner Funnels
Fitted specifically for Scienceware table-top Buchner funnels
GE Healthcare Whatman™ Quantitative Filter Paper: Grade 540 Circles Available on GSA/VA Contract for Federal Government customers only.
Use as a gravimetric filter during analysis of metals in acidic or alkaline solutions and in the collection of hydroxides. Whatman™ Quantitative Filter Paper: Grade 540 Circles acid-treated surface prevents tearing during filtration and allows the user to scrape precipitates from the paper.
GE Healthcare Whatman™ Quantitative Filter Paper: Grade 589/2 ½ Pleated Circles
Retains medium fine precipitates using material with virtually no ash content. Whatman™Quantitative Filter Paper: Grade 589/2 ½ Pleated Circles are suitable for a variety of routine quantitative analyses including analysis of aqueous suspension in paper, flour analysis, and sand content quantification.
GE Healthcare Whatman™ Grade 1573-1/2 Qualitative Filter Papers
A medium fast filter paper with high wet strength. This paper has the same chemical resistance characteristics as Grade 1573
GE Healthcare Whatman™ Qualitative Filter Papers - Prepleated Grade 113V
Savings in time required to quadrant-fold circles to fit conical filter funnels in repetitive or multiple analyses
GE Healthcare Whatman™ Qualitative Filter Paper: Grade 597½ Pleated Circles Available on GSA/VA Contract for Federal Government customers only.
Pleated to save time and achieve optimal fit in conical filter funnels. Whatman™ Qualitative Filter Paper: Grade 597½ Pleated Circles provide extra surface area to increase total loading capacity.
Fisherbrand™ Qualitative Grade Plain Filter Paper Circles and Sheets - P2 Grade
Ideal for clarifying and removing particles of 1μm and larger, and in applications that do not require a low-ash or ashless paper
Fisherbrand™ Qualitative Filter Paper: Grade 6 Circles
Filters particles twice as fast as Qualitative Filter Paper Grade 5. Fisherbrand™ Qualitative Grade 6 Filters provide fine particle retention for boiler water analysis.
Eisco Qualitative Filter Paper, 18cm
Student quality filter paper for use in schools or home. Diameter 18cm
Eisco Qualitative Filter Paper, 9cm
Student quality filter paper for use in schools or home.
GE Healthcare Whatman™ Reeve Angel 202 Creped Qualitative Circles
For routine, noncritical laboratory filtration applications such as general chemistry, pharmacology, emulsions, petroleum testing, soaps and detergents
  spinner