Glass Filtering Funnels
PYREX™ Buchner Funnels with Fritted Discs Available on GSA/VA Contract for Federal Government customers only.
Useful where filter paper would be attacked by material being filtered
DWK Life Sciences Kimble™ KIMAX™ Buchner Kimflow™ Fritted Disc Funnels Available on GSA/VA Contract for Federal Government customers only.
Use where filtrate would attack paper filters and membranes
DWK Life Sciences Kimble™ Kontes™ Parts for Microfiltration Assemblies
For use with ULTRA-WARE™ microfiltration assembly
Synthware™ Buchner Funnels with Perforated Glass Plate
With sealed in perforated glass disc to support filter media
PYREX™ Hirsch-Type Funnel with Fritted Disc
Ideal when necessary to wash precipitates and redissolve with chemicals that would attack filter paper
Synthware™ Buchner Funnels with Inner Joint
For use with filtration techniques
Synthware™ Buchner Filter Funnels
Used to filter particulates from substance
Synthware™ Addition Funnels with Pressure Equalizing Arm and Glass Stopcock
Cylindrical style addition funnel with pressure equalizing arm and glass stopcock
Eisco Funnel Filter 3 in. - Borosilicate Glass
Eisco Labs Borosilicate glass Funnel Filter 3 in. Diameter, Pitch of 60° angle. Long stem 2.75 in. One Funnel
Kontes™ Membrane Filtration Glassware from Restek
Used for routine filtration of corrosive liquids and removal of particles from HPLC solvents
Synthware™ Addition Funnels with Pressure Equalizing Arm and 1:5 PTFE Stopcock
Cylindrical style addition funnel with pressure equalizing arm and PTFE stopcock
Synthware™ Buchner Funnels with Inner Joint and Perforated Glass Plate
With sealed in perforated glass disc to support filter media
Synthware™ Addition Funnels with Pressure Equalizing Arm and PTFE Stopcock
Cylindrical style addition funnel with pressure equalizing arm and PTFE Stopcock
Synthware™ Addition Funnels with Pressure Equalizing Arm
Cylindrical style addition funnel with pressure equalizing arm
DWK Life Sciences Kimble™ Kontes™ Fritted Hirsch Filter Funnels
Directs filtrate through a reduced area frit to concentrate crystal residue for rewashing
Eisco™ Heavy Duty Glass Filter Funnels
Eisco™ Heavy Duty Glass Filter Funnels feature a thick wall design for enhanced durability.
DWK Life Sciences DURAN™ Filter Funnels
Mate to filtering flask via conical rubber seal (GUKO). Thermal shock and chemical resistant.
Synthware™ Pressure Equalizing Funnels with High Vacuum Valve
With extended tip valve for semi-needle valve adjustment
Synthware™ Filter Funnels with Fritted Disc
For introduction of powder or liquids into reaction systems
Synthware™ Filter Funnels with Fritted Disc and Glass Stopcock
Used for filtration followed by recrystallization on opposite side of disc of air sensitive compounds
Eisco™ Glass Funnel Filters Products for Science Education
Eisco™ Glass Funnel Filters are made from high quality borosilicate glass and designed with an angled stem and beaded rim for durability.
GSC Go Science Crazy Sintered Glass Funnels Products for Science Education
Quality borosilicate glass funnel with porous filter plate.
PYREX™ Hirsch Type, Fritted Disc Products for Science Education
Very useful in preparation work.
Eisco™ Glass Buchner Funnels with Jointed Stems Products for Science Education
Glass Buchner funnels with jointed stems and G-3 sintered disks. Made from high quality borosilicate 3.3 glass.
  spinner