Discs Filtration Membranes
EMD Millipore™ Silver Membrane Filters Available on GSA/VA Contract for Federal Government customers only.
Excellent chemical and temperature resistance
EMD Millipore™ MF-Millipore™ Mixed Cellulose Ester Membranes with Grid: 0.45μm Pore Size Available on GSA/VA Contract for Federal Government customers only.
Designed for a wide range of analytical and research applications. EMD Millipore MF-Millipore™ Mixed Cellulose Ester Membranes, 0.45µm pore size are available in a range of diameters. Composed of mixed cellulose membrane in black or white with a plain or gridded surface.
EMD Millipore™ Durapore™ PVDF Membrane Filters: 0.45μ Pore Size Available on GSA/VA Contract for Federal Government customers only.
Recommended when maximum recovery is important. EMD Millipore Durapore™ PVDF Membranes: 0.45μ Pore Size provide high flow rates and throughput, low extractables, broad chemical compatibility and low protein binding. Available in diameters ranging from 13mm to 142mm.
EMD Millipore™ Isopore™ Polycarbonate Membrane Filters Available on GSA/VA Contract for Federal Government customers only.
Use for analyses of airborne contaminants and other particles using optical or electron microscopy. EMD Millipore Isopore™ Polycarbonate Membrane Filters have a smooth, glass-like surface for clearer sample observation. Available in a variety of pore sizes/flow rates, diameters and colors.
EMD Millipore™ Durapore™ PVDF Membrane Filters: 0.22μ Pore Size Available on GSA/VA Contract for Federal Government customers only.
Recommended when maximum recovery is important. EMD Millipore Durapore™ PVDF Membranes: 0.22μ Pore Size provide high flow rates and throughput, low extractables, broad chemical compatibility and low protein binding. Available in diameters ranging from 13mm to 142mm.
Fisherbrand™ General Filtration Membrane Filters Available on GSA/VA Contract for Federal Government customers only.
Useful for most filter applications
EMD Millipore™ MF-Millipore™ Mixed Cellulose Ester Membranes: 0.22μm Pore Size Available on GSA/VA Contract for Federal Government customers only.
Designed for a wide range of analytical and research applications. EMD Millipore MF-Millipore™ Mixed Cellulose Ester Membranes, 0.22µm pore size are available in a range of diameters. Composed of plain, white mixed cellulose membrane.
EMD Millipore™ Express™ PLUS Membrane Filters Available on GSA/VA Contract for Federal Government customers only.
Plain white polyethersulfone membranes provide ultrafast filtration for a variety of applications
GE Healthcare Whatman™ Anodisc™ Filter Membranes Available on GSA/VA Contract for Federal Government customers only.
Constructed with precision pore structure and narrow pore size distribution. Whatman™ Anodisc™ Filter Membranes ensure a high level of particle removal efficiency. Captures microorganisms and particulates on the surface of the membrane for subsequent analysis.
GE Healthcare Whatman™ Nylon Membranes Available on GSA/VA Contract for Federal Government customers only.
Flexible and durable for easier handling.   GE Healthcare Nylon Membranes are naturally hydrophilic, eliminating need for wetting agents and prewetting when filtering aqueous solutions.
EMD Millipore™ Nylon Hydrophilic Membrane Filters Available on GSA/VA Contract for Federal Government customers only.
Filtration of aqueous or organic solutions
EMD Millipore™ MF-Millipore™ Mixed Cellulose Ester Membranes: 5.0μm Pore Size Available on GSA/VA Contract for Federal Government customers only.
Designed for a wide range of analytical and research applications. EMD Millipore MF-Millipore™ Mixed Cellulose Ester Membranes, 5.0µm pore size are available in a range of diameters. Composed of plain, white mixed cellulose membrane.
EMD Millipore™ Fluoropore™ PTFE Membrane Filter, Dia.: 47mm
Features broad chemical compatiblity with acids, bases and solvents.
GE Healthcare Whatman™ Type WCN Cellulose Nitrate Membranes Available on GSA/VA Contract for Federal Government customers only.
Recommended for majority of routine applications, this membrane is manufactured under strictly controlled conditions.  Whatman™ Type WCN Cellulose Nitrate Membranes narrow pore size distribution for improved surface capture and analysis.
GE Healthcare Whatman™ 47mm Nuclepore™ Polycarbonate Track-Etched Membranes Available on GSA/VA Contract for Federal Government customers only.
Manufactured from high-quality polycarbonate film and track-etched for high pore size precision. Whatman™ 47mm Nuclepore™ Polycarbonate Hydrophilic Membranes combine high flow rates and chemical and thermal resistance with sharply defined pores, making them suitable for microscopy, cytology, parasitology, and certain air and oceanography analyses.
EMD Millipore™ Fluoropore™ PTFE Membrane Filters Available on GSA/VA Contract for Federal Government customers only.
Hydrophobic membrane with laminated backing for easier handling
EMD Millipore™ MF-Millipore™ Mixed Cellulose Ester Membranes: 0.80μm Pore Size Available on GSA/VA Contract for Federal Government customers only.
Designed for air monitoring, bioassays, dairy microbiology, particle monitoring and particle removal. EMD Millipore MF-Millipore™ Mixed Cellulose Ester Membranes, 0.80µm pore size are available in a range of diameters. Composed of mixed cellulose membrane in black or white with a plain or gridded surface.
Sartorius™ Microsart™ @Filter 100 in Trays
Membrane filters suitable for use in the Microsart @Filter units
EMD Millipore™ Mitex™ PTFE Membrane Filters Available on GSA/VA Contract for Federal Government customers only.
Unlaminated for extreme chemical or temperature conditions
GE Healthcare Whatman™ PVP-Free Polycarbonate Cell Culture Membrane Available on GSA/VA Contract for Federal Government customers only.
Combines PVP-free polycarbonate with sharp, uniform cylindrical pores. Whatman™ PVP-Free Polycarbonate Cell Culture Membranes achieves a delicate balance between cell adhesion and migration for effective chemotaxis.
EMD Millipore™ Omnipore™ Membrane Filter Available on GSA/VA Contract for Federal Government customers only.
Compatible with virtually all solvents, acids and alkaline solutions
Sartorius™ Ultrafiltration Membrane Discs Available on GSA/VA Contract for Federal Government customers only.
For sample concentration in stirred cells
GVS Magna™ Nylon Membrane Filters
Plain white nylon membranes offer low extractables, high uniformity
Sartorius™ Microsart™ @Filter 250 in Trays Available on GSA/VA Contract for Federal Government customers only.
Membrane filters suitable for use in the Microsart @Filter units
EMD Millipore™ MF-Millipore™ Mixed Cellulose Ester Membranes: 3.0μm Pore Size Available on GSA/VA Contract for Federal Government customers only.
For use in air monitoring, fluid monitoring, particle collection and analysis, and QC of fluid holding tanks
EMD Millipore™ MF-Millipore™ Mixed Cellulose Ester Membranes: 8.0μm Pore Size Available on GSA/VA Contract for Federal Government customers only.
For use in Air monitoring, Fluid monitoring, particle collection and analysis, and QC of fluid holding tanks
  spinner