Motor Blowers
Thermo Scientific™ Blower Assembly Available on GSA/VA Contract for Federal Government customers only.
Permits exact balancing of room ventilating system with hood exhaust volume
Labconco™ Coated Steel Blower Products for Science Education
For low to moderately corrosive applications. Labconco™ Coated Steel Blower specifically designed to meet the demanding day-to-day ventilation requirements for wide range of hood choices.
Labconco™ PVC Motor Blower Products for Science Education
Labconco™ PVC Blower provide optimum chemical resistance and are made specifically to handle high order oxidizers such as perchloric acid.
Labconco™ Motor Blowers: Coated Steel
For low to moderately corrosive applications
Labconco™ Intelli-Sense™ Multi-Speed Fiberglass Blowers
Intelli-Sense Multi-Speed Blowers are weatherized, direct drive, non-explosion-proof blowers controlled by two or three position switch
Labconco™ Fiberglass Motor Blowers
Ideal for fume hood exhaust systems in moderate to highly corrosive conditions
Labconco™ Intelli-Sense™ Multi-Speed Coated Steel Blowers: 230V
Programmable up to three different speeds, including night set-back option
Labconco™ Intelli-Sense™ Multi-Speed PVC Blowers
PVC Blowers provide optimum chemical resistance and are made specifically to handle high order oxidizers such as perchloric acid
Labconco™ Intelli-Sense™ Multi-Speed Coated Steel Blowers: 115V
Programmable up to three different speeds, including night set-back option
Labconco™ PVC Motor Blowers
Handle extremely corrosive atmospheres produced by high order oxidizers such as perchloric acid
Dolly for Extract-All™ Series 987 Compact Air Cleaner
For easy transport of series 987 Compact Air Cleaner
Noise Silencer for Extract-All™ Series 982 Motor Blowers
For quieter operation of high pressure Extract-All motor blowers
Extract-All™ Series 982 Motor Blowers
Compact high-pressure blowers designed to accommodate multiple extractor arms
Airclean™ System Polypropylene Hood Blowers
Used especially in situations that require resistance to chemical agents, silent operation and energy savings
  spinner