Specialty Hoods and Enclosures
Thermo Scientific™ Microm™ Air Filtration System
Features a small footprint and three speed settings for ease of use
Fisher Scientific™ PCR Workstation
Fisher Scientific PCR Workstation provides ISO Class 5 conditions appropriate for PCR experiments.
Bel-Art™ SP Scienceware™ Techni-Dome™ 360 Balance Enclosure
Offers generous space for safely positioning an analytical balance, in a virtually draft-free and compact environment.
Labconco™ RXPert™ Single Filtered Balance Systems
The patented RXPert Filtered Balance System is Class I Enclosure that meets USP <800> requirements for non-sterile compounding when ducted. Models feature an all-in-one factory-installed canopy and damper for thimble ducting to the outside.
Labconco™ XPert™ Balance Enclosures, Standard Height: 60 in.W
Protects user by keeping powders, particulates and fumes contained
Labconco™ Protector™ Stainless-Steel Radioisotope Hoods, Deep
Provide protection from applications using radiochemicals
Labconco™ XPert™ Balance Enclosures - Tall, 5 Ft. Wide
Enclosures protect user by keeping powders, particulates and fumes contained
Labconco™ XPert™ Balance Enclosures - Tall, 6 Ft. Wide
Enclosures protect user by keeping powders, particulates and fumes contained
Labconco™ XPert™ Filtered Balance Station, Deep: 6ft., 115V 60Hz
Built-in HEPA filter, mounted in the upper plenum, removes particulates and exhausts clean air to the atmosphere
Labconco™ RXPert™ Double Filtered Balance Systems
The patented RXPert Filtered Balance System is Class I Enclosure that meets USP <800> requirements for non-sterile compounding when ducted. Models feature 2 HEPA filters to provide redundant HEPA filtration in series and therefore require NO ductwork.
Labconco™ XPert™ Nano Enclosures: 5ft. Width Without Built-in Ionizer
Keep hazardous powders and particulates contained during procedures
Labconco™ Protector™ Stainless-Steel Radioisotope Hood
Provide protection from applications using radiochemicals with a Labconco Protector™ Stainless-Steel Radioisotope Hood.
Labconco™ XPert™ Filtered Balance Station, Deep: 5ft., 230V 50/60Hz
Built-in HEPA filter, mounted in the upper plenum, removes particulates and exhausts clean air to the atmosphere
Labconco™ XPert™ Bulk Powder Filtered Systems
Protects from hazardous airborne particulates and fumes generated during powder transfer operations
Labconco™ XPert™ Balance Enclosures - Tall, 2 Ft. Wide
Enclosures protect user by keeping powders, particulates and fumes contained
Labconco™ XPert™ Balance Enclosures - Tall, 3 Ft. Wide
Enclosures protect user by keeping powders, particulates and fumes contained
Labconco™ XPert™ Filtered Balance Station, Deep: 6ft., 230V 50/60Hz
Built-in HEPA filter, mounted in the upper plenum, removes particulates and exhausts clean air to the atmosphere
Labconco™ FilterMate™ Portable Exhausters Products for Science Education
The FilterMate uses HEPA filter, carbon filter or combination of both filters to remove hazardous powders, particulates or vapors from exhaust air stream and return filtered air to environment.
Labconco™ XPert™ Balance Enclosures — Tall and Deep, 6 ft. Wide
Enclosures protect user by keeping powders, particulates and fumes contained
Labconco™ CApture™ Portable Fuming System
Quickly fixes latent fingerprints at a crime scene using microburst or traditional CA fuming methods, while protecting the user from hazardous fumes
Labconco™ Protector™ Downdraft Powder Station
The Labconco Protector™ Downdraft Powder Station keeps users safe by containing powders during fingerprint processing.
Labconco™ XPert™ Balance Enclosures, Standard Height: 72 in.W
Protects user by keeping powders, particulates and fumes contained
Labconco™ XPert™ Balance Enclosure, Standard Height: 36 in.W
Protect users by keeping powders, particulates and fumes contained by using a standing height Labconco XPert™ Balance Enclosure.
Labconco™ XPert™ Easy-Order Balance Enclosure Systems
Low-profile enclosure systems provide user protection by containing powders, particulates, and fumes during weighing procedures.
Labconco™ XPert™ Nano Enclosures: 5ft. Width With Built-in Ionizer
Keep hazardous powders and particulates contained during procedures
Labconco™ XPert™ Balance Enclosures, Standard Height: 96 in.W
Protects user by keeping powders, particulates and fumes contained
Labconco™ XPert™ Bulk Powder Enclosures
Exhausts hazardous airborne particulates and fumes to outside
Labconco™ Purifier™ Filtered PCR Enclosure
Labconco Purifier™ Filtered PCR Enclosure protects work from particulate contamination.
Labconco™ XPert™ Filtered Balance Station: Nominal Width, 6ft.
Protects user during powder weighing operations
  spinner