HEPA Filtered Enclosures
Labconco™ RXPert™ Single Filtered Balance Systems
The patented RXPert Filtered Balance System is Class I Enclosure that meets USP <800> requirements for non-sterile compounding when ducted. Models feature an all-in-one factory-installed canopy and damper for thimble ducting to the outside.
Labconco™ RXPert™ Double Filtered Balance Systems
The patented RXPert Filtered Balance System is Class I Enclosure that meets USP <800> requirements for non-sterile compounding when ducted. Models feature 2 HEPA filters to provide redundant HEPA filtration in series and therefore require NO ductwork.
Labconco™ XPert™ Bulk Powder Filtered Systems
Protects from hazardous airborne particulates and fumes generated during powder transfer operations
Labconco™ Protector™ Downdraft Powder Station
The Labconco Protector™ Downdraft Powder Station keeps users safe by containing powders during fingerprint processing.
Labconco™ XPert™ Bulk Powder Enclosures
Exhausts hazardous airborne particulates and fumes to outside
Labconco™ XPert™ Bulk Powder Filtered Stations
Designed to protect the user during powder transfer operations
Labconco™ XPert™ Filtered Balance System with Guardian 1000 Digital Airflow Monitor
With the Labconco XPert™ Filtered Balance System with Guardian 1000 Digital Airflow Monitor you can provide user protection by keeping powders and particulates contained during weighing procedures.
Labconco™ XPert™ Filtered Balance Systems without Airflow Monitor, 230V
Keeps powders and particulates contained during procedures by using the Labconco XPert™ Filtered Balance Systems without Airflow Monitor.
Labconco™ XPert™ Filtered Balance Systems with Guardian™ Airflow Monitor
Labconco XPert™ Filtered Balance Systems come with Guardian™ Airflow Monitor for added convenience.
Labconco™ HEPA Filter for Rotary Vane Vacuum Pump and FreeZone™ Freeze Dry Systems
Fits between freeze dryer and pump to prevent entry of particulate contaminants into pump
AirClean™ Systems HEPA Filters
High efficiency particulate air (HEPA) filter, AirClean™ Systems HEPA Filters is used with Ductless fume hoods and drying cabinets.
AirClean™ Systems PowderSafe™ Type A Ductless Balance Enclosure
Based on SafeChange technology that minimizes threat of exposure to toxic powders during pre-filter replacement
Airclean™ System TEE Probe Storage Cabinets
Designed to accommodate up to three disinfected probes
  spinner