Chemical Resistant Gloves
Ansell™ MICROFLEX™ MegaPro™ Gloves
Ansell™ MICROFLEX™ MegaPro™ Gloves offer advanced protection and durability for demanding jobs
Ansell™ TouchNTuff™ Sterile Polyisoprene Gloves
Minimizes the risk of latex sensitization.
Ansell™ NeoTouch™ Neoprene Gloves, 9 in.
Comfortable, synthetic single-use gloves
Ansell™ Touch N Tuff™ Disposable Nitrile Gloves, Powder-free
Proven splash resistance against hazardous chemicals. Ansell Edmont Touch N Tuff™ Disposable Nitrile Gloves provide a comfortable fit and minimize hand fatigue.
Ansell™ AlphaTec™ Butyl Glove 38-514
The thinnest butyl glove providing the best resistance to the most aggressive chemicals with dexterity and comfort.
Ansell TouchNTuff™ DermaShield™ 73-711
Ideal glove for workers in aseptic environments who are concerned about allergy risks.
Microflex™ Cobalt™ X Nitrile Gloves
Extra thickness for additional protection
Kimberly-Clark Professional™ KleenGuard™ G29 Solvent Gloves
Provide the dexterity of thin mil gloves with greater durability for superior protection. Kimberly Clark™ Professional KleenGuard™ G29 Solvent Gloves feature neoprene blend construction to protect against solvents and won't break apart after 15 minutes of work like thin mil gloves.
Ansell™ AlphaTec™ 87-208 Orange Heavyweight Latex Gloves
Superior tensile strength and sensitivity to resist acids, ketones, salts, detergents, alcohols, alkalis and fats
Ansell™ AlphaTec™ 88-394 Latex Gloves
A combination of great protection and a high level of dexterity, makes this glove Ideally suited for the food processing industry
Ansell™ Canners and Handlers RBC Latex Gloves
Medium-duty gloves feature rolled beaded cuff and diamond embossed grip
Microflex™ 93-850 Ultimate Barrier Nitrile Disposable Gloves
Allows workers in demanding environments to experience higher levels of protection using gloves that exceed the highest known standard for barrier integrity. Significantly reduces the risk of harmful exposure.
Honeywell™ North™ Curved Hand Butyl Gloves
Curved hand design for better dexterity
Honeywell™ North™ Butyl Gloves
High permeation resistance to gas or water vapors
Ansell™ TouchNTuff™ Disposable Chemical Resistant Nitrile Gloves
Proven splash resistance against hazardous chemicals.
SHOWA™ Master™ Natural Rubber Gloves
Unsupported natural rubber for tactile sensitivity, exceptional tensile strength, and tear and puncture resistance
Fisherbrand™ Double-Dipped PVC Gloves: Gauntlet Length
Provide enhanced cut resistance and excellent wet or slippery grip
Ansell™ BioClean Suprene™ Sterile Neoprene Cleanroom Gloves
Neoprene provides added strength and enhanced softness. Nitritex™ BioClean Suprene™ Sterile Neoprene Cleanroom Gloves are resistant to a range of chemicals, designed for double-donning, have antistatic properties and feature a hand-specific shape for a comfortable and ergonomic fit.
Honeywell™ CSM Glovebox Ambidextrous Gloves
Provides resistance to ozone and oxidizing chemicals such as alcohols, alkalis and acids.
Ansell™ ChemTek™ Viton/Butyl Chemical Handling Gloves
Offering the ultimate in splash and immersion protection.
Ansell™ AlphaTec™ 19-024 Neoprene Dipped Gloves
Neoprene-dipped coating on a double insulating liner for improved resistance in hot conditions
SHOWA™ Viton™ and Viton II Gloves
Provide highest chemical-resistance barrier to aromatic hydrocarbons such as benzene, toluene, or xylene.
Ansell™ AlphaTec™ 88-392 Latex Gloves
Unlined industrial latex gloves
Bel-Art™ SP Scienceware™ Glove Box Economy Sleeved Gloves
Compatible with Scienceware Portable Glove Box or any glove box with 6 in. glove ports
Guardian Manufacturing 8B1532A, Ambidextrous 15 mil Thick Butyl Glovebox Gloves
Manufactured with a unique solution-dipped process, providing unparalleled protection for users. Guardian™ Ambidextrous 15 mil Thick Butyl Glovebox Gloves provide accurate controls every step of the way in order to attain the necessary strength.
Ansell™ Sol-Knit™ Gloves
Offers chemical resistance of nitrile with the strength and comfort of cotton
Guardian Manufacturing 8B1532, Hand Specific 15 mil Butyl Glovebox Gloves
Manufactured with a unique solution-dipped process, providing unparalleled protection for users. Guardian™ Hand Specific 15 mil Thick Butyl Glovebox Gloves provide accurate controls every step of the way in order to attain the necessary strength.
MAPA™ Blue-Grip™ Latex Gloves
Provides easy on-off thermal protection
Guardian Manufacturing 8B3032, Hand Specific 30 mil Butyl Glovebox Gloves
Manufactured with a unique solution-dipped process, providing unparalleled protection for users. Guardian™ Hand Specific 30 mil Thick Butyl Glovebox Gloves provide accurate controls every step of the way in order to attain the necessary strength.
  spinner