Black Stains Dyes and Indicators
Eriochrome™ Black T Solution, Fisher Chemical
Chemical Name or Material: Eriochrome™ Black T Solution CAS: 67-56-1 CAS: 1787-61-7 CAS: 1336-21-6 CAS: 7732-18-5 MDL Number: MFCD00003935 Synonym: Mordant Black 11
Eriochrome™ Black T (Certified), Fisher Chemical
Chemical Name or Material: Eriochrome™ Black T CAS: 1787-61-7 Purity Grade: Certified Grade Molecular Formula: C20H12N3NaO7S Formula Weight: 461.38 MDL Number: MFCD00003935 Synonym: Superchrome Black TS, 1-(1-Hydroxy-2-naphthylazo)-6-nitro-2-naphthol-4-sulfonic Acid Sodium Salt, Mordant Black 11
Sudan Black B, high purity biological stain, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sudan Black B, high purity biological stain CAS: 4197-25-5 Purity Grade: High Purity Biological Stain Molecular Formula: C29H24N6 Formula Weight: 456.54 MDL Number: MFCD00006919 Synonym: C.I. 26150, Fat Black HB, Solvent Black 3
Eriochrome™Black T, ACS reagent, indicator grade, ACROS Organics™
Chemical Name or Material: Eriochrome™ Black T CAS: 1787-61-7 Purity Grade: ACS Reagent Grade Molecular Formula: C20H12N3NaO7S Formula Weight: 461.38 Synonym: C.I. 14645, 3-Hydroxy-4-(1-hydroxy-2-naphthylazo)-7-nitro-1-naphthalene sulfonic acid, sodium salt, Mordant Black 11
Eriochrome™ Black T, pure, indicator grade, ACROS Organics™
Chemical Name or Material: Eriochrome™ Black T CAS: 1787-61-7 Purity Grade: Pure Molecular Formula: C20H12N3NaO7S Formula Weight: 461.38 MDL Number: MFCD00003935 Synonym: C.I. 14645, 3-Hydroxy-4-(1-hydroxy-2-naphthylazo)-7-nitro-1-naphthalene sulfonic acid, sodium salt, Mordant Black 11
Eriochrome Black TS, Ricca Chemical
Chemical Name or Material: Eriochrome Black Name Note: Test Solution CAS: 67-56-1 CAS Min %: 93.0 CAS Max %: 97.0 CAS: 5470-11-1 CAS Min %: 4.72 CAS Max %: 4.92 CAS: 1787-61-7 CAS Min %: 0.47 CAS Max %: 0.49 Physical Form: Liquid
Starch Indicator, Aqueous Solution, 0.5% (w/v), Ricca Chemical
CAS: 9005-84-9 Physical Form: Liquid Assay: 0.50% Assay Percent Range: 0.5% (w/v)
Eriochrome Black T Indicator, 1% (w/w) in Sodium Chloride, Ricca Chemical
Chemical Name or Material: Eriochrome Black T Indicator CAS: 1787-61-7 CAS: 7647-14-5 Physical Form: Powder
Sudan Black, Fisher BioReagents™
Chemical Name or Material: Sudan Black CAS: 4197-25-5 Molecular Formula: C29H24N6 Formula Weight: 456.54
Mordant Black 11 Triturate R, 1% (w/w) in Sodium Chloride, Ricca Chemical
Chemical Name or Material: Mordant Black 11 Triturate Name Note: Reagent CAS: 7647-14-5 CAS Min %: 97.0 CAS Max %: 100.0 CAS: 1787-61-7 CAS Min %: 0.98 CAS Max %: 1.02 Physical Form: Powder
Water Hardness Indicator, 0.5% (w/v) Eriochrome Black T in 2-Methoxyethanol, Ricca Chemical
Chemical Name or Material: Water Hardness Indicator Name Note: 0.5% (w/v) Eriochrome Black T in 2-Methoxyethanol CAS: 109-86-4 CAS Min %: 98.0 CAS Max %: 100.0 CAS: 1787-61-7 CAS Min %: 0.47 CAS Max %: 0.49 Physical Form: Liquid
Eriochrome Black T Solution, ACROS Organics™
Chemical Name or Material: Eriochrome Black T solution CAS: 1787-61-7 Molecular Formula: C20H12N3NaO7S Formula Weight: 461.38 MDL Number: MFCD00003935 Synonym: C.I. 14645, 3-Hydroxy-4-(1-hydroxy-2-naphthylazo)-7-nitro-1-naphthalene sulfonic acid, sodium salt, Mordant Black 11
Calcon, pure, ACROS Organics™
Chemical Name or Material: Calcon™ CAS: 2538-85-4 Purity Grade: Pure Molecular Formula: C20H13N2NaO5S Formula Weight: 416.39 MDL Number: MFCD00004069 Synonym: C.I. 15705, Eriochrome™ Blue Black R, C.I. 15705, Mordant Black 17, Mordant Black 17, Zinchrome R
Sudan Black B MP Biomedicals
Chemical Name or Material: Sudan Black B CAS: 4197-25-5 Molecular Formula: C29H24N6 Formula Weight: 456.5 Synonym: Solvent Black 3
Amido Black 10B, Fisher BioReagents
Chemical Name or Material: Amido Black 10B CAS: 1064-48-8 Molecular Formula: C22 H14N6Na2O9S2 Formula Weight: 616.5 Synonym: Naphthol Blue Black
Eriochrome Black T ACS Reagent Grade, Ricca Chemical
Chemical Name or Material: Eriochrome Black T Name Note: Certified CAS: 1787-61-7 CAS Min %: 98.0 CAS Max %: 100.0 Purity Grade: ACS Reagent Grade
Sudan Black B, MP Biomedicals
Chemical Name or Material: Sudan Black B CAS: 4197-25-5 Formula Weight: 456.55
Chlorazol Black E, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Chlorazol Black E Name Note: Pure CAS: 1937-37-7 Purity Grade: Pure Molecular Formula: C34H25N9Na2O7S2 Formula Weight: 781.73 MDL Number: MFCD00066363
Sudan Black B, certified; ACROS Organics™
Chemical Name or Material: Sudan Black B, certified Name Note: certified CAS: 4197-25-5 Molecular Formula: C29H24N6 Formula Weight: 456.54 MDL Number: MFCD00006919 Synonym: C.I. 26150, Fat Black HB, Solvent Black 3
Diazine Black, MP Biomedicals
Chemical Name or Material: Diazine Black CAS: 4443-99-6
Brilliant Black, MP Biomedicals
Chemical Name or Material: Brilliant Black CAS: 2519-30-4 Formula Weight: 871.74
Brilliant Black BN, MP Biomedicals™
Chemical Name or Material: Brilliant Black BN CAS: 2519-30-4 Physical Form: Powder Molecular Formula: C28H17N5O14S4Na4 Formula Weight: 867.7 Synonym: CI 28440, Food Black 1
Sudan Black B, Powder, HARLECO™, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sudan Black B, Powder Name Note: HARLECO™ Purity Grade: Certified Biological Stain Physical Form: Powder Synonym: Solvent Black 3
Amido black 10 B (C.I.20470), For Electrophoresis, Certistain™, EMD Millipore™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Amido black 10 B (C.I.20470) CAS: 1064-48-8 Molecular Formula: C22H14N6Na2O9S2 Formula Weight: 616.49 Synonym: Black acid 1, Naphthol blue black
Direct Black 22, ACROS Organics™
Chemical Name or Material: Direct Black 22 CAS: 6473-13-8 Molecular Formula: C44H32N13Na3O11S3 Formula Weight: 1083.98 Synonym: C.I. 35435
Fast Sulphon Black F, ACROS Organics™
Chemical Name or Material: Fast Sulphon Black F CAS: 3682-47-1 Molecular Formula: C30H17N4Na3O11S3 Formula Weight: 774.63 MDL Number: MFCD00021612 Synonym: Acid Black 32, C.I. 26990
  spinner