Red Stains Dyes and Indicators
Eosin Y (Certified Biological Stain), Fisher Chemical
CAS: 17372-87-1 Molecular Formula: C20H12Br4Na2O8 Molecular Weight (g/mol): 745.904 MDL Number: MFCD00005040 InChI Key: MASXMTNVNASWNH-UHFFFAOYSA-L Synonym: D&C Red No. 22, Acid Red 87 PubChem CID: 91886399 IUPAC Name: disodium;2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate;trihydrate SMILES: C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)Br)[O-])Br)Br)Br)C(=O)[O-].O.O.O.[Na+].[Na+]
Phenol Red, Free Acid (Certified ACS), Fisher Chemical
CAS: 143-74-8 Molecular Formula: C19H14O5S Molecular Weight (g/mol): 354.376 MDL Number: MFCD00003552 InChI Key: BELBBZDIHDAJOR-UHFFFAOYSA-N Synonym: 4,4'-(3H-2,1-Benzoxathiol-3-yildene)bisphenol S,S-Dioxide, 4, 4'-(3H-2, 1-Benzoxathiol-3-yildene)bisphenol S PubChem CID: 4766 ChEBI: CHEBI:31991 IUPAC Name: 4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol SMILES: C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)O
Nile Red 99%, ACROS Organics™
CAS: 7385-67-3 Molecular Formula: C20H18N2O2 Molecular Weight (g/mol): 318.376 MDL Number: MFCD00011639 InChI Key: VOFUROIFQGPCGE-UHFFFAOYSA-N Synonym: Nile blue A oxazone PubChem CID: 65182 ChEBI: CHEBI:52169 IUPAC Name: 9-(diethylamino)benzo[a]phenoxazin-5-one SMILES: CCN(CC)C1=CC2=C(C=C1)N=C3C4=CC=CC=C4C(=O)C=C3O2
Eosin Y (1% Solution in Water), Fisher Chemical
CAS: 17372-87-1 Molecular Formula: C20H12Br4Na2O8 Molecular Weight (g/mol): 745.904 MDL Number: MFCD00005040 InChI Key: MASXMTNVNASWNH-UHFFFAOYSA-L PubChem CID: 91886399 IUPAC Name: disodium;2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate;trihydrate SMILES: C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)Br)[O-])Br)Br)Br)C(=O)[O-].O.O.O.[Na+].[Na+]
Fuchsin Basic (Certified Biological Stain), Fisher Chemical
CAS: 632-99-5 Molecular Formula: C20H20ClN3 Molecular Weight (g/mol): 337.851 MDL Number: MFCD00001657 InChI Key: AXDJCCTWPBKUKL-UHFFFAOYSA-N Synonym: Magenta, Pararosaniline PubChem CID: 12447 IUPAC Name: 4-[(4-aminophenyl)-(4-imino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]aniline;hydrochloride SMILES: CC1=CC(=C(C2=CC=C(C=C2)N)C3=CC=C(C=C3)N)C=CC1=N.Cl
Sudan IV (Certified Biological Stain), Fisher Chemical
CAS: 85-83-6 Molecular Formula: C24H20N4O Molecular Weight (g/mol): 380.451 MDL Number: MFCD00003893 InChI Key: KMDLOETUWUPGMB-UHFFFAOYSA-N Synonym: Scarlet Red, Lipid Crimson, Solvent Red 24, Oil Red PubChem CID: 6797604 IUPAC Name: 1-[[2-methyl-4-[(2-methylphenyl)diazenyl]phenyl]hydrazinylidene]naphthalen-2-one SMILES: CC1=CC=CC=C1N=NC2=CC(=C(C=C2)NN=C3C(=O)C=CC4=CC=CC=C43)C
Alizarin Red S, pure, certified, ACROS Organics™
CAS: 130-22-3 Molecular Formula: C14H7NaO7S Molecular Weight (g/mol): 342.253 MDL Number: MFCD00013049 InChI Key: HFVAFDPGUJEFBQ-UHFFFAOYSA-M Synonym: 9,10-Dihydro-3,4-dihydroxy-9,10-dioxo-2-anthracenesulfonic acid, sodium salt, 9, 10-Dihydro-3, 4-dihydroxy-9, Mordant Red 3, Sodium alizarinesulfonate PubChem CID: 3955344 ChEBI: CHEBI:87358 IUPAC Name: sodium;3,4-dihydroxy-9,10-dioxoanthracene-2-sulfonate SMILES: C1=CC=C2C(=C1)C(=O)C3=CC(=C(C(=C3C2=O)O)O)S(=O)(=O)[O-].[Na+]
Methyl Red, Sodium Salt (Certified ACS), Fisher Chemical
CAS: 845-10-3 Molecular Formula: C15H14N3NaO2 Molecular Weight (g/mol): 291.286 MDL Number: MFCD00002426 InChI Key: GNTPCYMJCJNRQB-UHFFFAOYSA-M Synonym: Acid Red 2, Sodium Salt PubChem CID: 4465632 IUPAC Name: sodium;2-[[4-(dimethylamino)phenyl]diazenyl]benzoate SMILES: CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)[O-].[Na+]
Neutral Red (Certified Biological Stain), Fisher Chemical
CAS: 553-24-2 Molecular Formula: C15H17ClN4 Molecular Weight (g/mol): 288.779 MDL Number: MFCD00012651 InChI Key: PGSADBUBUOPOJS-UHFFFAOYSA-N Synonym: Basic Red 5, Toluylene Red PubChem CID: 11105 ChEBI: CHEBI:86370 IUPAC Name: 8-N,8-N,3-trimethylphenazine-2,8-diamine;hydrochloride SMILES: CC1=CC2=NC3=C(C=C(C=C3)N(C)C)N=C2C=C1N.Cl
Phenol Red, Sodium Salt (Certified ACS), Fisher Chemical
CAS: 34487-61-1 Molecular Formula: C19H13NaO5S Molecular Weight (g/mol): 376.358 MDL Number: MFCD00066901 InChI Key: HKHYOKBQJILTEI-UHFFFAOYSA-M Synonym: Phenolsulfonephthalein Sodium Salt PubChem CID: 23686673 IUPAC Name: sodium;4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenolate SMILES: C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)[O-].[Na+]
Nile Red Fluorescent stain for intracellular lipid droplets. MP Biomedicals
CAS: 7385-67-3 Molecular Formula: C20H18N2O2 Molecular Weight (g/mol): 318.376 InChI Key: VOFUROIFQGPCGE-UHFFFAOYSA-N Synonym: Nile Blue A Oxazone, 9-diethylamino-5H-benzo-alpha-phenoxazine-5-one PubChem CID: 65182 ChEBI: CHEBI:52169 IUPAC Name: 9-(diethylamino)benzo[a]phenoxazin-5-one SMILES: CCN(CC)C1=CC2=C(C=C1)N=C3C4=CC=CC=C4C(=O)C=C3O2
Congo Red (Certified Biological Stain), Fisher Chemical
CAS: 573-58-0 Molecular Formula: C32H22N6Na2O6S2 Molecular Weight (g/mol): 696.664 InChI Key: IQFVPQOLBLOTPF-UHFFFAOYSA-L Synonym: Diphenyl Bisazonaphthionic Acid, Direct Red 28 PubChem CID: 11313 ChEBI: CHEBI:34653 IUPAC Name: disodium;4-amino-3-[[4-[4-[(1-amino-4-sulfonatonaphthalen-2-yl)diazenyl]phenyl]phenyl]diazenyl]naphthalene-1-sulfonate SMILES: C1=CC=C2C(=C1)C(=CC(=C2N)N=NC3=CC=C(C=C3)C4=CC=C(C=C4)N=NC5=C(C6=CC=CC=C6C(=C5)S(=O)(=O)[O-])N)S(=O)(=O)[O-].[Na+].[Na+]
Erythrosin B, pure, certified, ACROS Organics™
CAS: 16423-68-0 Molecular Formula: C20H6I4Na2O5 Molecular Weight (g/mol): 879.86 MDL Number: MFCD00144257 InChI Key: RAGZEDHHTPQLAI-UHFFFAOYSA-L Synonym: Acid Red 51, C.I. 45430 PubChem CID: 12961638 IUPAC Name: disodium;2',4',5',7'-tetraiodo-3-oxospiro[2-benzofuran-1,9'-xanthene]-3',6'-diolate SMILES: C1=CC=C2C(=C1)C(=O)OC23C4=CC(=C(C(=C4OC5=C(C(=C(C=C35)I)[O-])I)I)[O-])I.[Na+].[Na+]
Neutral Red, MP Biomedicals
1:300 Solution Storage temperature: 15-30°C
Nile red, MP Biomedicals™
CAS: 7385-67-3 Molecular Formula: C20H18N2O2 Molecular Weight (g/mol): 318.376 MDL Number: MFCD00011639 InChI Key: VOFUROIFQGPCGE-UHFFFAOYSA-N Synonym: Nile Blue A Oxazone, 9-Diethylamino-5H-benzo[a]phenoxazine-5-one PubChem CID: 65182 ChEBI: CHEBI:52169 IUPAC Name: 9-(diethylamino)benzo[a]phenoxazin-5-one SMILES: CCN(CC)C1=CC2=C(C=C1)N=C3C4=CC=CC=C4C(=O)C=C3O2
Methyl Red, Free Acid (Certified), Fisher Chemical
CAS: 493-52-7 Molecular Formula: C15H15N3O2 Molecular Weight (g/mol): 269.304 MDL Number: MFCD00002425 InChI Key: CEQFOVLGLXCDCX-UHFFFAOYSA-N Synonym: Acid Red 2, o-(p-Dimethylaminophenylazo)benzoic Acid, Acid Red 2 PubChem CID: 10303 IUPAC Name: 2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid SMILES: CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O
Nuclear Fast Red, 94%, pure, ACROS Organics™
CAS: 6409-77-4 Molecular Formula: C14H11NNaO7S Molecular Weight (g/mol): 360.292 MDL Number: MFCD00078493 InChI Key: KDEATZKCHKVXGA-UHFFFAOYSA-N Synonym: Calcium Red, C.I. 60760 PubChem CID: 131675027 IUPAC Name: 4-amino-1,3-dihydroxy-9,10-dioxoanthracene-2-sulfonic acid;molecular hydrogen;sodium SMILES: [HH].C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=C(C(=C3N)O)S(=O)(=O)O)O.[Na]
Nuclear fast red (C.I. 60760), For Microscopy, Certistain™, EMD Millipore™
CAS: 6409-77-4 Molecular Formula: C14H11NNaO7S Molecular Weight (g/mol): 360.292 InChI Key: KDEATZKCHKVXGA-UHFFFAOYSA-N Synonym: C.I. 60760 PubChem CID: 131675027 IUPAC Name: 4-amino-1,3-dihydroxy-9,10-dioxoanthracene-2-sulfonic acid;molecular hydrogen;sodium SMILES: [HH].C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=C(C(=C3N)O)S(=O)(=O)O)O.[Na]
Congo Red, Indicator grade, ACROS Organics™
CAS: 573-58-0 Molecular Formula: C32H22N6Na2O6S2 Molecular Weight (g/mol): 696.664 MDL Number: MFCD00004028 InChI Key: IQFVPQOLBLOTPF-UHFFFAOYSA-L Synonym: C.I. 22120, Direct Red 28 PubChem CID: 11313 ChEBI: CHEBI:34653 IUPAC Name: disodium;4-amino-3-[[4-[4-[(1-amino-4-sulfonatonaphthalen-2-yl)diazenyl]phenyl]phenyl]diazenyl]naphthalene-1-sulfonate SMILES: C1=CC=C2C(=C1)C(=CC(=C2N)N=NC3=CC=C(C=C3)C4=CC=C(C=C4)N=NC5=C(C6=CC=CC=C6C(=C5)S(=O)(=O)[O-])N)S(=O)(=O)[O-].[Na+].[Na+]
EOSIN Y, Ricca Chemical
CAS: 17372-87-1 Molecular Formula: C20H12Br4Na2O8 Molecular Weight (g/mol): 745.904 InChI Key: MASXMTNVNASWNH-UHFFFAOYSA-L PubChem CID: 91886399 IUPAC Name: disodium;2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate;trihydrate SMILES: C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)Br)[O-])Br)Br)Br)C(=O)[O-].O.O.O.[Na+].[Na+]
Ruthenium Red tetrahydrate, pure, ACROS Organics™
CAS: 12790-48-6 Molecular Formula: Cl6H36N14O6Ru3-14 Molecular Weight (g/mol): 844.29 MDL Number: MFCD00167176 InChI Key: IMVKVBRYNZXYAP-UHFFFAOYSA-H PubChem CID: 108195 IUPAC Name: azanide;oxygen(2-);ruthenium;ruthenium(5+);hexachloride;tetrahydrate SMILES: [NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].O.O.O.O.[O-2].[O-2].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ru].[Ru+5].[Ru+5]
Phenol Red, ACROS Organics™
CAS: 143-74-8 Molecular Formula: C19H14O5S Molecular Weight (g/mol): 354.376 MDL Number: MFCD00003552 InChI Key: BELBBZDIHDAJOR-UHFFFAOYSA-N Synonym: Phenolsulfonephthalein PubChem CID: 4766 ChEBI: CHEBI:31991 IUPAC Name: 4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol SMILES: C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)O
Eosin Y, pure, Indicator grade, ACROS Organics™
CAS: 17372-87-1 Molecular Formula: C20H12Br4Na2O8 Molecular Weight (g/mol): 745.904 MDL Number: MFCD00005040 InChI Key: MASXMTNVNASWNH-UHFFFAOYSA-L Synonym: 2';,4';,5';,7';-Tetrabromofluorescein, disodium salt, Eosin Yellowish, 2';, 4';, 5'; PubChem CID: 91886399 IUPAC Name: disodium;2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate;trihydrate SMILES: C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)Br)[O-])Br)Br)Br)C(=O)[O-].O.O.O.[Na+].[Na+]
Direct Red 80, Alfa Aesar™
CAS: 2610-10-8 Molecular Formula: C45H26N10Na6O21S6 Molecular Weight (g/mol): 1373.05 MDL Number: MFCD00054389 InChI Key: LDCKXVAPQFUIOI-ZFLDVXHKSA-H PubChem CID: 57369623 IUPAC Name: hexasodium;4-oxo-7-[[5-oxo-7-sulfonato-6-[[2-sulfonato-4-[(4-sulfonatophenyl)diazenyl]phenyl]hydrazinylidene]naphthalen-2-yl]carbamoylamino]-3-[[2-sulfonato-4-[(4-sulfonatophenyl)diazenyl]phenyl]hydrazinylidene]naphthalene-2-sulfonate SMILES: C1=CC(=CC=C1N=NC2=CC(=C(C=C2)NN=C3C(=CC4=C(C3=O)C=CC(=C4)NC(=O)NC5=CC6=C(C=C5)C(=O)C(=NNC7=C(C=C(C=C7)N=NC8=CC=C(C=C8)S(=O)(=O)[O-])S(=O)(=O)[O-])C(=C6)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+]
Methyl Red hydrochloride, ACS reagent, ACROS Organics™
CAS: 63451-28-5 Molecular Formula: C15H16ClN3O2 Molecular Weight (g/mol): 305.762 MDL Number: MFCD00012614 InChI Key: ICTWKXAVJAHRMK-UHFFFAOYSA-N Synonym: Acid Red 2, 2-[4-(Dimethylamino)phenylazo]benzoic acid hydrochloride, C.I. 13020, C.I. 13020 PubChem CID: 113227 IUPAC Name: 2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid;hydrochloride SMILES: CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O.Cl
Erythrosin B, Alfa Aesar™
CAS: 568-63-8 Molecular Formula: C40H14I8Na2O10 Molecular Weight (g/mol): 1715.757 MDL Number: MFCD00144257 InChI Key: DKRYETBOLGDIOK-UHFFFAOYSA-L PubChem CID: 131676154 IUPAC Name: disodium;2-(3-hydroxy-2,4,5,7-tetraiodo-6-oxoxanthen-9-yl)benzoic acid;2-(2,4,5,7-tetraiodo-3-oxido-6-oxoxanthen-9-yl)benzoate SMILES: C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)I)O)I)I)I)C(=O)O.C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)I)[O-])I)I)I)C(=O)[O-].[Na+].[Na+]