Platforms and Clamps for Baths
Thermo Scientific™ Clips and Fasteners for Precision™ Water Baths Available on GSA/VA Contract for Federal Government customers only.
Use these clips and fasteners Thermo Scientific™ Precision Reciprocal, Dubnoff and Shallow-Form Shaking Water Baths.
Fisherbrand™ Water Bath Clamps
Holds glass tubes and thermometers onto walls of glass water bath.
Eppendorf™ Universal Platform for Model C76 Water Bath Shaker Available on GSA/VA Contract for Federal Government customers only.
Both universal platforms and dedicated platforms are available in a wide variety of material and size perfect for virtually every requirement. Universal platforms have multiple holes allowing you to use virtually any combination of clamps and other accessories. Dedicate platforms are used when just one flask size will be used.
  spinner