Circulating Baths
Thermo Scientific™ Precision™ Circulating Water Baths Available on GSA/VA Contract for Federal Government customers only.
Achieve outstanding temperature uniformity for your samples with perimeter-directed water flow on Precision circulating water baths. Featuring advanced temperature controller.
Thermo Scientific™ ARCTIC A40 Refrigerated Circulator Available on GSA/VA Contract for Federal Government customers only.
A40 refrigerated bath circulator provides efficient cooling for open and closed applications, with temperatures ranging from −40° to +200°C and 12L maximum bath volume.
Thermo Scientific™ ARCTIC A25 Refrigerated Circulators Available on GSA/VA Contract for Federal Government customers only.
A25 refrigerated bath circulator provides efficient cooling for open and closed applications, with temperatures ranging from −25° to +200°C and 12L maximum bath volume.
Thermo Scientific™ Precision™ Circulating Water Baths
Achieve outstanding temperature uniformity for your samples with perimeter-directed water flow on Precision circulating water baths. Featuring advanced temperature controller.
Thermo Scientific™ SAHARA S7 Stainless-Steel Heated Bath Circulators Available on GSA/VA Contract for Federal Government customers only.
Perfect for internal or external circulation. Offered in a wide selection for efficient heating from ambient 13° to 300°C, with maximum bath volume of 8 liters.
Thermo Scientific™ ARCTIC A10 Refrigerated Circulators Available on GSA/VA Contract for Federal Government customers only.
A10 refrigerated bath circulators provide efficient cooling for open and closed applications, with temperatures ranging from −10° to +100°C and 6L maximum bath volume.
Thermo Scientific™ SAHARA PPO S21P Heated Bath Circulators Available on GSA/VA Contract for Federal Government customers only.
Ideal when visibility of your application is required. These baths maintain temperatures from ambient +13° to 100°C, with maximum bath volume of 21 liters.
Fisherbrand™ Isotemp™ Heated Bath Circulators: Model 5150 H7 Available on GSA/VA Contract for Federal Government customers only.
Reliable, high-wattage heaters that deliver rapid fluid heat up and ensure fluid compatibility for your application.
Fisherbrand™ IsoTemp™ Refrigerated/Heated Bath Circulator Basic Starter Kit
For limited time, bundle and save up to $900 on select Fisher Scientific™ Isotemp™ Refrigerated/Heated Bath Circulator packages (Models 4100 R20, 6200 R28 and 6200 R35)
Thermo Scientific™ SAHARA S19T Transparent Acrylic Heated Bath Available on GSA/VA Contract for Federal Government customers only.
Ideal when visibility of your application is required
Fisherbrand™ Isotemp™ Heated Bath Circulators: Model 6200 H24 Available on GSA/VA Contract for Federal Government customers only.
Reliable, high-wattage heaters that deliver rapid fluid heat up and ensure fluid compatibility for your application.
Thermo Scientific™ SAHARA S5P Heated PPO Bath Available on GSA/VA Contract for Federal Government customers only.
A polyphenylene oxide bath is an economical alternative to stainless steel
Fisherbrand™ Isotemp™ Heated Bath Circulators: Model 4100 H11 Available on GSA/VA Contract for Federal Government customers only.
Reliable, high-wattage heaters that deliver rapid fluid heat up and ensure fluid compatibility for your application
Fisherbrand™ Isotemp™ Heated Bath Circulators: Model 4100 H7 Available on GSA/VA Contract for Federal Government customers only.
Reliable, high-wattage heaters that deliver rapid fluid heat up and ensure fluid compatibility for your application.
Thermo Scientific™ ARCTIC A25B Open Refrigeration Bath Steel Available on GSA/VA Contract for Federal Government customers only.
For use with ARCTIC A25B Open Refrigeration Bath Circulator
Thermo Scientific™ ARCTIC A40 Refrigeration Bath Steel Available on GSA/VA Contract for Federal Government customers only.
Designed to ensure accuracy and reproducibility of your liquid temperature control procedures
Thermo Scientific™ SAHARA S45SS Stainless Steel Heated Bath Available on GSA/VA Contract for Federal Government customers only.
Durable, seamless stainless steel baths for high temperature applications
Thermo Scientific™ SAHARA S7SS Stainless Steel Heated Bath Available on GSA/VA Contract for Federal Government customers only.
Durable, seamless stainless steel bath for high temperature applications
Thermo Scientific™ SAHARA Acrylic S12T Heated Bath Circulators Available on GSA/VA Contract for Federal Government customers only.
Ideal when visibility of your application is required, with efficient heating from ambient plus 13°C to a maximum of 80°C.
Thermo Scientific™ SAHARA S13 Stainless-Steel Heated Bath Circulators Available on GSA/VA Contract for Federal Government customers only.
Perfect for internal or external circulation. Offered in a wide selection for efficient heating from ambient 13° to 300°C, with maximum bath volume of 12 liters.
Thermo Scientific™ ARCTIC A10B Refrigerated Circulators Available on GSA/VA Contract for Federal Government customers only.
A10B refrigerated bath features a large work area, enabling high throughput and workflow efficiency, with temperatures ranging from −10° to +100°C and 30L maximum bath volume.
Thermo Scientific™ SAHARA S49SS Stainless Steel Heated Bath Available on GSA/VA Contract for Federal Government customers only.
Durable, seamless stainless steel bath for high temperature applications
Thermo Scientific™ Ultra-Low Temperature Circulators Available on GSA/VA Contract for Federal Government customers only.
These Ultra-Low Temperature Circulators meet a variety of application needs. The ULT baths are designed primarily to circulate to external closed loop applications.
Thermo Scientific™ SAHARA S21 Stainless-Steel Heated Bath Circulators Available on GSA/VA Contract for Federal Government customers only.
Perfect for internal or external circulation. Offered in a wide selection for efficient heating from ambient 13° to 200°C, with maximum bath volume of 19 liters.
Thermo Scientific™ SAHARA SC150L S45 Heated Stainless Steel Bath Circulators Available on GSA/VA Contract for Federal Government customers only.
Rugged and corrosion-resistant for high temperature applications up to 300°C
Thermo Scientific™ SAHARA Heated PPO Bath Circulators Available on GSA/VA Contract for Federal Government customers only.
These polyphenylene oxide baths are an economical alternative to stainless steel
Fisherbrand™ Isotemp™ Heated Bath Circulators: Model 5150 H24
Reliable, high-wattage heaters that deliver rapid fluid heat up and ensure fluid compatibility for your application.
Thermo Scientific™ SAHARA S21P Heated PPO Bath Available on GSA/VA Contract for Federal Government customers only.
An economical alternative to stainless steel that delivers exceptional temperature performance
  spinner