Heated Circulating Baths
Thermo Scientific™ SAHARA S7 Stainless-Steel Heated Bath Circulators Available on GSA/VA Contract for Federal Government customers only.
Perfect for internal or external circulation. Offered in a wide selection for efficient heating from ambient 13° to 300°C, with maximum bath volume of 8 liters.
Thermo Scientific™ SAHARA S13 Stainless-Steel Heated Bath Circulators Available on GSA/VA Contract for Federal Government customers only.
Perfect for internal or external circulation. Offered in a wide selection for efficient heating from ambient 13° to 300°C, with maximum bath volume of 12 liters.
Fisherbrand™ Isotemp™ Heated Bath Circulators: Model 6200 H24 Available on GSA/VA Contract for Federal Government customers only.
Reliable, high-wattage heaters that deliver rapid fluid heat up and ensure fluid compatibility for your application.
Thermo Scientific™ SAHARA PPO S21P Heated Bath Circulators Available on GSA/VA Contract for Federal Government customers only.
Ideal when visibility of your application is required. These baths maintain temperatures from ambient +13° to 100°C, with maximum bath volume of 21 liters.
Thermo Scientific™ SAHARA Acrylic S19T Heated Bath Circulators Available on GSA/VA Contract for Federal Government customers only.
Ideal when visibility of your application is required, with efficient heating from ambient plus 13°C to a maximum of 80°C.
Thermo Scientific™ SAHARA S6T Transparent Acrylic Heated Bath Available on GSA/VA Contract for Federal Government customers only.
Ideal when visibility of your application is required
Fisherbrand™ Isotemp™ Heated Bath Circulators: Model 4100 H11 Available on GSA/VA Contract for Federal Government customers only.
Reliable, high-wattage heaters that deliver rapid fluid heat up and ensure fluid compatibility for your application
Fisherbrand™ Isotemp™ Heated Bath Circulators: Model 5150 H7 Available on GSA/VA Contract for Federal Government customers only.
Reliable, high-wattage heaters that deliver rapid fluid heat up and ensure fluid compatibility for your application.
Fisherbrand™ Isotemp™ Heated Bath Circulators: Model 4100 H7 Available on GSA/VA Contract for Federal Government customers only.
Reliable, high-wattage heaters that deliver rapid fluid heat up and ensure fluid compatibility for your application.
Thermo Scientific™ SAHARA S19T Transparent Acrylic Heated Bath Available on GSA/VA Contract for Federal Government customers only.
Ideal when visibility of your application is required
Thermo Scientific™ SAHARA AC 150-S21 Stainless Steel Heated Bath Circulators Available on GSA/VA Contract for Federal Government customers only.
Rugged and corrosion-resistant for high temperature applications up to 300°C.
Thermo Scientific™ SAHARA S45 Stainless-Steel Heated Bath Circulators Available on GSA/VA Contract for Federal Government customers only.
Perfect for internal or external circulation. Offered in a wide selection for efficient heating from ambient 13° to 200°C, with maximum bath volume of 41 liters.
Thermo Scientific™ SAHARA S21P Heated PPO Bath Available on GSA/VA Contract for Federal Government customers only.
An economical alternative to stainless steel that delivers exceptional temperature performance
Thermo Scientific™ SAHARA SC150 S14P Heated PPO Bath Circulators Available on GSA/VA Contract for Federal Government customers only.
Polyphenylene oxide baths are an economical alternative to stainless steel and thermally resistant up to 100°C
Fisherbrand™ Isotemp™ 4100 H12A Open Heated Acrylic Bath Circulators Available on GSA/VA Contract for Federal Government customers only.
Acrylic open heated baths are fitted with bridge plates for placement of the control head
Thermo Scientific™ SAHARA Heated PPO Bath Circulators Available on GSA/VA Contract for Federal Government customers only.
These polyphenylene oxide baths are an economical alternative to stainless steel
Thermo Scientific™ SAHARA SC150L S45 Heated Stainless Steel Bath Circulators Available on GSA/VA Contract for Federal Government customers only.
Rugged and corrosion-resistant for high temperature applications up to 300°C
Thermo Scientific™ SAHARA S21 Stainless-Steel Heated Bath Circulators Available on GSA/VA Contract for Federal Government customers only.
Perfect for internal or external circulation. Offered in a wide selection for efficient heating from ambient 13° to 200°C, with maximum bath volume of 19 liters.
Thermo Scientific™ SAHARA S13 Stainless Steel Heating Bath Available on GSA/VA Contract for Federal Government customers only.
Durable, seamless stainless steel baths for high temperature applications
Thermo Scientific™ SAHARA S7SS Stainless Steel Heated Bath Available on GSA/VA Contract for Federal Government customers only.
Durable, seamless stainless steel bath for high temperature applications
Thermo Scientific™ SAHARA PPO S14P Heated Bath Circulators Available on GSA/VA Contract for Federal Government customers only.
Ideal when visibility of your application is required. These baths maintain temperatures from ambient +13° to 100°C, with maximum bath volume of 14 liters.
Thermo Scientific™ SAHARA Acrylic S6T Heated Bath Circulators Available on GSA/VA Contract for Federal Government customers only.
Ideal when visibility of your application is required, with efficient heating from ambient plus 13°C to a maximum of 80°C.
Thermo Scientific™ SAHARA S21SS Stainless Steel Heated Bath Available on GSA/VA Contract for Federal Government customers only.
Durable, seamless stainless steel bath for high temperature applications
Thermo Scientific™ SAHARA AC 150-S49 Heated Stainless Steel Bath Circulator Available on GSA/VA Contract for Federal Government customers only.
Rugged and corrosion-resistant for high temperature applications up to 300°C.
Thermo Scientific™ SAHARA S49SS Stainless Steel Heated Bath Available on GSA/VA Contract for Federal Government customers only.
Durable, seamless stainless steel bath for high temperature applications
Thermo Scientific™ SAHARA SC150 S45 Heated Bath Circulators Available on GSA/VA Contract for Federal Government customers only.
Rugged and corrosion-resistant for high temperature applications up to 300°C
Thermo Scientific™ SAHARA S49 Stainless-Steel Heated Bath Circulators Available on GSA/VA Contract for Federal Government customers only.
Perfect for internal or external circulation. Offered in a wide selection for efficient heating from ambient 13° to 200°C, with maximum bath volume of 53 liters.
Thermo Scientific™ SAHARA AC200 S14P Heated PPO Bath Circulator Available on GSA/VA Contract for Federal Government customers only.
Polyphenylene oxide baths an economical alternative to stainless steel
  spinner