Viscosity Bath
Cannon™ IBF Bath Oil for Glass Capillary Viscometer
Ideal bath oil for Cannon heated baths with applications at ambient termperature and above
Cannon™ CT-1000 Constant-Temperature Viscosity Bath
For precise viscosity determinations with glass capillary viscometers.
Cannon™ CT-500 Constant-Temperature Bath
Maintains temperatures ±0.01°C between 20° and 100°C, meeting ASTM D 445 precision specification for kinematic viscosity measurements using glass capillary viscometers
Koehler™ Instrument KV1000 Digital Constant Temperature Kinematic Viscosity Bath
Constant temperature bath for kinematic viscosity testing of petroleum products
Koehler™ Instrument KV5000 Kinematic Viscosity Bath
Provides automatic viscosity measurements of petroleum and petrochemical products
Koehler™ Instrument SV3000 Saybolt Viscosity Bath
Determines the time required for 60mL of sample to flow through a calibrated orifice under precisely controlled conditions
Fungilab™ Thermocap Plus Kinematic Water Bath
Increase the presicion and temperature control of a capillary viscometer. Thermocap Plus Kinematic Water Bath is ideal for Cannon-Fenske and Ubbelohde capillary viscometers.
  spinner