Alcohol Burners
DWK Life Sciences Wheaton™ Alcohol Burner Available on GSA/VA Contract for Federal Government customers only.
Ideal for microscopy applications
GSC Go Science Crazy Alcohol Burner with Dual Wicks
A great addition to any lab in which a small burner is needed. Made of durable metal to prevent breakage. The GSC Alcohol Burner with Dual Wicks is an all metal, permanent wick alcohol burner with a capacity 100ml tank. The wick portion is a ceramic fiber inserted into a thin copper tube. This construction allows for constant flame. The burner is fueled by high quality alcohol which can be found in most local hardware stores.
Fisherbrand™ Alcohol Burner Products for Science Education
For use with any experiment requiring low heat.
Fisherbrand™ Brass Alcohol Burner Products for Science Education
Polished metal alcohol burner is much safer than glass burners.
Supertek Scientific Aluminium Burner Products for Science Education
Methylated spirit burner with wick, wick holder and cap.
Supertek Scientific Wickless Alcohol Burners Products for Science Education
Safe alternative to conventional alcohol burners
  spinner